Recommended Products
manufacturer/tradename
NIST®
packaging
pkg of (5x10g)
application(s)
food and beverages
format
mixture
General description
Other Notes
Reference/Informational value is provided for the following analytes.
Daidzin (C21H20O9), Genistin (C15H10O5), Glycitin (C22H22O10), Total protein
See certificate for values and more details at nist.gov/SRM.
Daidzin (C21H20O9), Genistin (C15H10O5), Glycitin (C22H22O10), Total protein
See certificate for values and more details at nist.gov/SRM.
Legal Information
NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service