Skip to Content
Merck
All Photos(2)

Key Documents

71478

Supelco

Nα-(2,4-Dinitro-5-fluorophenyl)-L-alaninamide

for chiral derivatization, LiChropur, ≥99.0%

Synonym(s):

FDAA, Marfey’s reagent

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9FN4O5
CAS Number:
Molecular Weight:
272.19
Beilstein:
6820069
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.22

grade

for chiral derivatization

Quality Level

Assay

≥99.0% (sum of enantiomers, TLC)
≥99.0%

form

powder

optical activity

[α]20/D +56±2°, c = 1% in acetone

optical purity

enantiomeric ratio: ≥99.5:0.5 (HPLC)

quality

LiChropur

technique(s)

HPLC: suitable

storage temp.

2-8°C

SMILES string

C[C@H](Nc1cc(F)c(cc1[N+]([O-])=O)[N+]([O-])=O)C(N)=O

InChI

1S/C9H9FN4O5/c1-4(9(11)15)12-6-2-5(10)7(13(16)17)3-8(6)14(18)19/h2-4,12H,1H3,(H2,11,15)/t4-/m0/s1

InChI key

NEPLBHLFDJOJGP-BYPYZUCNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nα-(2,4-Dinitro-5-fluorophenyl)-L-alaninamide (FDAA) is a chiral derivatizing agent (CDA), has high enantioselectivity but low sensitivity as compared to other CDAs. It is generally used to assign the stereochemistry of amino acids in trace amounts.

Application

FDAA was used as derivatizing reagent, in a study performed to understand unusual amino acids using reversed phase high performance liquid chromatography-electrospray ionization mass spectrometry (RPHPLC-ESI-MS).

Other Notes

Derivatization reagent for the assay of unusual chiral α-amino acid analogs

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chirality determination of unusual amino acids using precolumn derivatization and liquid chromatography-electrospray ionization mass spectrometry.
Hess S
Journal of Chromatography A, 1035(2), 211-219 (2004)
R Bhushan et al.
Amino acids, 36(3), 571-579 (2008-07-10)
Some non-protein alpha-amino acids were derivatized with 1-fluoro-2,4-dinitrophenyl-5-L-alaninamide (Marfey's reagent, MR, FDNP-L-Ala-NH(2),) and four of its structural variants FDNP-L-Phe-NH(2), FDNP-L-Val-NH(2), FDNP-L-Leu-NH(2) and FDNP-L-Pro-NH(2). The resultant diastereomers were separated by normal and reversed phase thin layer chromatography (TLC) and reversed phase
Michael J Berna et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 846(1-2), 359-363 (2006-09-12)
The ability to selectively measure serine enantiomer concentrations in rat brain microdialysate is essential during drug discovery to study the interaction of d-serine with the N-methyl-d-aspartate (NMDA) subtype of the glutamate receptor. NMDA receptor-stimulating agents, such as d-serine, have been
G Szókán et al.
Journal of chromatography, 444, 115-122 (1988-07-01)
Reversed-phase high-performance liquid chromatography and pre-column derivatization with 1-fluoro-2,4-dinitrophenyl-5-L-alanine amide (Marfey's reagent) were used for monitoring racemization in peptides, amino acids and their derivatives by separation of optical isomers of amino acids. The technique was applied to the analysis of
Sonja Hess et al.
Analytical biochemistry, 311(1), 19-26 (2002-11-21)
Silk fibroins from moth larvae and spiders are composed of highly repetitive Ala- and Gly-rich blocks that determine their structure, properties, and function. To investigate the metabolic integration of isotopically labeled amino acids in the excreted silk, the enrichment of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service