16259
Silicon Standard for AAS
TraceCERT®, 1 g/L Si in NaOH (nominal concentration)
Synonym(s):
Silicon AAS Standard Solution
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116107
NACRES:
NA.24
Recommended Products
grade
certified reference material
TraceCERT®
Quality Level
product line
TraceCERT®
concentration
1 g/L Si in NaOH (nominal concentration)
technique(s)
AAS: suitable
application(s)
industrial qc
pharmaceutical
format
single component solution
General description
This certified reference material (CRM) is produced in accordance with ISO 17034 and characterized in accordance with ISO/IEC 17025. This CRM is traceable to SI unit kg and measured against primary material from a National Metrology Institute (NMI), e.g. NIST.
Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.
Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.
Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.
Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.
Application
Silicon standard may be used as calibration standard during determination of silicon from powdered aluminium oxide and petrochemical products using graphite-furnace atomic-absorption spectrometry (GF-AAS).
Analysis Note
1 g/L Si in 2% sodium hydroxide, prepared with high purity silicon metal, NaOH and water
Other Notes
For a complete product listing of our TraceCERT® range of CRMs for ICP and AAS, technical information, and example certificates please visit our ICP & AAS standards website
Legal Information
TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
H Minami et al.
Fresenius' journal of analytical chemistry, 370(7), 855-859 (2001-09-25)
A direct method for determination of silicon in powdered high-purity aluminium oxide samples, by slurry sampling with in situ fusion graphite-furnace atomic-absorption spectrometry (GF-AAS), has been established. A slurry sample was prepared by 10-min ultrasonication of a powdered sample in
Niobium carbide as permanent modifier for silicon determination in petrochemical products by emulsion-based sampling GF AAS.
Luz, Maciel S., and Pedro V. Oliveira
Fuel: The Science and Technology of Fuel and Energy, 255-260 (2014)
Niobium carbide as permanent modifier for silicon determination in petrochemical products by emulsion-based sampling GF AAS.
Luz, Maciel S., and Pedro V. Oliveira
Fuel: The Science and Technology of Fuel and Energy, 255-260 (2014)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service