Skip to Content
Merck
All Photos(1)

Key Documents

MAB5432

Sigma-Aldrich

Anti-CREB Antibody

Chemicon®, from mouse

Synonym(s):

cAMP-response Element Binding Protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

monoclonal

species reactivity

human, mouse, rat

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CREB1(1385)

General description

CREB is a beta ZIP transcription factor that activates target genes through cAMP response elements. CREB is able to mediate signals from numerous physiological stimuli, resulting in regulation of a broad array of cellular responses. While CREB is expressed in numerous tissues, it plays a large regulatory role in the nervous system. CREB is believed to play a key role in promoting neuronal survival, precursor proliferation, neurite outgrowth and neuronal differentiation in certain neuronal populations. CREB promotes outgrowth and differentiation as a mediator of Neurotrophin pathways. CREB is able to selectively activate numerous downstream genes through interactions with different dimerization partners. CREB is activated by phosphorylation at Ser133 by various signaling pathways including Erk, Ca2+ and stress signaling.

Specificity

Reacts with CREB.

Immunogen

Recombinant CREB-A protein.

Application

Anti-CREB Antibody is a Mouse Monoclonal Antibody for detection of CREB also known as cAMP-response Element Binding Protein & has been validated in ELISA & WB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors
Western blot: 1-5 μg/mL. Recognizes the ~43kDa protein.

ELISA: 0.1-1.0 μg/mL

Optimal working dilutions must be determined by end user.

Linkage

Replaces: 04-218

Physical form

Format: Purified
Purified immunoglobulin. Liquid in PBS containing 0.1% sodium azide.

Storage and Stability

Maintain at 2-8°C in undiluted aliquots for up to 6 months after date of receipt.

Analysis Note

Control
Jurkat nuclear extract, A-431 whole cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yan-Yan Sun et al.
Scientific reports, 6, 30743-30743 (2016-08-05)
The number and subtype composition of N-methyl-d-aspartate receptor (NMDAR) at synapses determines their functional properties and role in learning and memory. Genetically increased or decreased amount of GluN2B affects hippocampus-dependent memory in the adult brain. But in some experimental conditions
Hong Cao et al.
Neuroscience bulletin, 28(1), 77-87 (2012-01-12)
The rostral anterior cingulate cortex (rACC) is implicated in processing the emotional component of pain. N-methyl-D-aspartate receptors (NMDARs) are highly expressed in the rACC and mediate pain-related affect by activating a signaling pathway that involves cyclic adenosine monophosphate (cAMP)/protein kinase
C E M Fernandes et al.
Brazilian journal of medical and biological research = Revista brasileira de pesquisas medicas e biologicas, 52(5), e8334-e8334 (2019-05-01)
Studies have shown that an injection with the histamine H4 receptor agonist VUF-8430 modulates emotional memory processes. In the present study, the aim was to verify if intraperitoneal (ip) injection of VUF-8430 (500 ng/kg) in mice affects the synthesis of
Wakako Kawarazaki et al.
Journal of the American Society of Nephrology : JASN, 23(6), 997-1007 (2012-03-24)
Experiments with hyperaldosteronemic animals suggest that, despite lowering plasma aldosterone, salt worsens renal injury by paradoxical activation of the mineralocorticoid receptor (MR). Salt and aldosterone synergistically contribute to renal impairment through Rac1-mediated activation of the MR, but whether angiotensin II
Xiao-Dan Han et al.
Molecular medicine reports, 11(1), 226-230 (2014-10-24)
Sevoflurane is an inhaled anesthetic that is widely used in clinical practice, particularly for pediatric anesthesia. Previous studies have suggested that sevoflurane may induce neurotoxicity in the brains of neonatal mice. In the present study, the possible mechanism of neurodegeneration

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service