Skip to Content
Merck
All Photos(1)

Key Documents

MAB3741

Sigma-Aldrich

Anti-Protein Tyrosine Phosphatase µ Antibody, clone SBK15

ascites fluid, clone SBK15, Chemicon®

Synonym(s):

Receptor-type Protein-tyrosine Phosphatase mu, R-PTP-MU

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

SBK15, monoclonal

species reactivity

rat, bovine, mouse, human, canine

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PTPRM(5797)

Specificity

Recognizes human PTPμ (Protein Tyrosine Phosphatase mu), a Receptor-type protein-tyrosine phosphatase. This antibody recognizes the cytoplasmic domain of PTPμ.

Immunogen

Corresponds to amino acids 774-1452 of human PTPμ, purified from sf9 cells.

Application

Research Category
Signaling
Research Sub Category
Kinases & Phosphatases
This Anti-Protein Tyrosine Phosphatase μ Antibody, clone SBK15 is validated for use in IC, IP, WB for the detection of Protein Tyrosine Phosphatase μ.
Western blot (1:1,000)

Immunoprecipitation

Immunostaining (1:200)

Optimal working dilutions must be determined by end user.

Physical form

Ascites fluid.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 12 months from date of receipt. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shanshan Zhang et al.
The American journal of pathology, 188(4), 995-1006 (2018-01-30)
Primary liver cancer consists mainly of hepatocellular carcinoma (HCC) and intrahepatic cholangiocarcinoma (ICC). A subset of human HCCs expresses a ICC-like gene signature and is classified as ICC-like HCC. The Hippo pathway is a critical regulator of normal and malignant
Keiko Takahashi et al.
PloS one, 12(5), e0177192-e0177192 (2017-05-26)
Renal vascular development is a coordinated process that requires ordered endothelial cell proliferation, migration, intercellular adhesion, and morphogenesis. In recent decades, studies have defined the pivotal role of endothelial receptor tyrosine kinases (RPTKs) in the development and maintenance of renal
Christian Griñan-Ferré et al.
Frontiers in aging neuroscience, 8, 241-241 (2016-11-03)
With the increase in life expectancy, aging and age-related cognitive impairments are becoming one of the most important issues for human health. At the same time, it has been shown that epigenetic mechanisms are emerging as universally important factors in
Laura Fernández-Sánchez et al.
Frontiers in cellular neuroscience, 9, 484-484 (2016-01-07)
Retinitis pigmentosa includes a group of progressive retinal degenerative diseases that affect the structure and function of photoreceptors. Secondarily to the loss of photoreceptors, there is a reduction in retinal vascularization, which seems to influence the cellular degenerative process. Retinal
Shuji Wakatsuki et al.
Genes to cells : devoted to molecular & cellular mechanisms, 19(1), 66-77 (2013-11-22)
After peripheral nerve injury, Schwann cells gain a migratory phenotype and remodel their extracellular matrix to provide a supportive environment for axonal regeneration. The soluble neuregulin-1 isoform, that is, glial growth factor (GGF), is expressed in regenerating axons of injured

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service