Z303860
Quickfit® wide-neck reaction flask
capacity 10,000 mL, Spherical
Synonym(s):
FR10LF
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
Recommended Products
material
borosilicate glass
round bottom
wide-neck
sterility
non-sterile
feature
closure type Lid
Spherical
packaging
pack of 1
manufacturer/tradename
Quickfit FR 10LF
availability
not available in USA and Canada
capacity
10,000 mL
Looking for similar products? Visit Product Comparison Guide
General description
Reaction flasks have a flat flange so that lids can be quickly and easily detached. Flanges have 100 mm bore for easy addition or removal of starting materials and products, as well as for simple cleaning. Z653322 has 75mm bore. Order lids and clips separately.
NOTE: Z338052 is cylindrical.
NOTE: Z338052 is cylindrical.
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service