Z303399
Quickfit® double surface (Davies) condenser
jacket L 205 mm, joint: ST/NS 29/32
Synonym(s):
C5/24/SC
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
Recommended Products
Joint
joint: ST/NS29/32
manufacturer/tradename
Quickfit C 5/24/SC
availability
not available in USA and Canada
not available in USA and Canada
jacket L
205 mm
surface area
300 cm2
General description
Increased surface area and effectiveness when compared with the corresponding Liebig condenser. Inner cooling surface gives a baffling effect, while outer cooling surface prevents creep and loss of vapour when used with low boiling liquids.
Legal Information
Quickfit is a registered trademark of SciLabware Limited