SYNF724250
Synthware™ Filtering flask with 10 mm OD hose connection
capacity 250 mL, neck joint: ST/NS 24/40
Synonym(s):
Synthware F724250, filtering flask with outer joint
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121804
NACRES:
NB.51
Recommended Products
material
flat bottom flask
glass flask
Joint
joint: ST/NS neck24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F724250
capacity
250 mL
Looking for similar products? Visit Product Comparison Guide
General description
Heavy wall design made from tubing to ensure excellent quality with 10 mm hose connection
Legal Information
Synthware is a trademark of Kemtech America Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service