SYNB257040
Synthware™ mineral oil bubbler
O.D. × H 185 mm
Synonym(s):
Mineral Oil Bubbler, Synthware B257040
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
Recommended Products
packaging
pkg of 1 ea
manufacturer/tradename
Synthware B257040
O.D. × H
185 mm
Looking for similar products? Visit Product Comparison Guide
General description
With 10 mm O.D. hose connection.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service