DWK63A53P
Kimble® Disposable Pasteur Pipet
L 5.75 in., Borosilicate glass/PTFE, Cotton-plugged
Synonym(s):
63A53P
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121500
NACRES:
NB.04
Recommended Products
material
Borosilicate glass/PTFE
feature
Cotton-plugged
packaging
pkg of 1000 ea
manufacturer/tradename
Kimble® 63A53P
L
5.75 in.
Looking for similar products? Visit Product Comparison Guide
General description
These Pasteur-type pipets are intended for one-time use in the transfer of small volumes.
Application
Bacteriology, immunology, hematology, and serology
Features and Benefits
Removes risk of cross-contamination of high value specimens and laboratory reagents.
Legal Information
KIMBLE is a registered trademark of DWK Life Sciences
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service