Skip to Content
Merck
All Photos(4)

Key Documents

739421

Sigma-Aldrich

Silver nanowires

diam. × L 50 nm × 6 μm, 0.5% (isopropyl alcohol suspension)

Synonym(s):

Silver nanofibers, Silver nanowhiskers, Silver nanowire

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ag
CAS Number:
Molecular Weight:
107.87
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

form

liquid (suspension)
nanowires

Quality Level

concentration

0.5% (isopropyl alcohol suspension)

diam. × L

50 nm × 6 μm

density

0.785 g/mL at 25 °C

SMILES string

[Ag]

InChI

1S/Ag

InChI key

BQCADISMDOOEFD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Silver nanowires (AgNWs) are 1-D silver based nanostructures which have a diameter of 60 nm and a 10 μm length. They can be synthesized by the reduction of silver nitrate (AgNO3) in presence of silver bromide (AgBr). They may also be prepared by the arc discharge mechanism of immersed silver electrodes.

Application

AgNWs have excellent electrical, optical and mechanical properties which make them a suitable material for flexible electronics based applications such as solar cells and film heaters.
Silver nanowires are useful in a wide variety of conductive, optical and anti-microbial applications such as touchscreen displays, medical imaging and sterile clothing.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

53.6 °F - closed cup

Flash Point(C)

12 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Silver nanowires: Synthesis technologies, growth mechanism and multifunctional applications
Zhang P, et al.
Materials Science and Engineering, B, 223(5), 1-23 (2017)
Crystalline silver nanowires by soft solution processing
Sun Y, et al.
Nano Letters, 2(2), 165-168 (2002)
Scalable coating and properties of transparent, flexible, silver nanowire electrodes
Hu L, et al.
ACS Nano, 4(5), 2955-2963 (2010)
Preparation of silver nanowires and their application in conducting polymer nanocomposites
Abbasi NM, et al.
Materials Chemistry and Physics, 166(5), 1-15 (2015)
Highly stretchable and sensitive strain sensor based on silver nanowire-elastomer nanocomposite
Amjadi M, et al.
ACS Nano, 8(5), 5154-5163 (2014)

Articles

The ability to pattern conductive electrodes is technologically relevant for several applications, including photovolatics, displays, sensors, and biomedical devices.

Silver nanomaterials have unique physical, chemical, and optical properties that are currently being leveraged for a wide variety of biological applications.

Among various ceramics, one-dimensional (1-D) piezoelectric ceramics have attracted significant scientific attention for use in energy harvesting.

TiO2 exhibits wide band gap semiconductor and memristor properties electronically, with high opacity and UV absorbance optically.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service