Skip to Content
Merck
All Photos(1)

Key Documents

247138

Sigma-Aldrich

Chlorocarbonylsulfenyl chloride

96%

Synonym(s):

(Chlorothio)formyl chloride, Chloroformylsulfenyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCOSCl
CAS Number:
Molecular Weight:
130.98
Beilstein:
506318
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

form

liquid

refractive index

n20/D 1.517 (lit.)

bp

98 °C (lit.)

density

1.552 g/mL at 25 °C (lit.)

functional group

chloro

storage temp.

2-8°C

SMILES string

ClSC(Cl)=O

InChI

1S/CCl2OS/c2-1(4)5-3

InChI key

MNOALXGAYUJNKX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Chlorocarbonylsulfenyl chloride has been used in the preparation of:
  • 5-(1,2,3,4-tetra-O-acetyl-alpha-D-xylopyranos-5S-C-yl)-1,3,4-oxathiazol-2-one
  • fluorinated oxathialones
  • polyfluoroalkylchlorothioformates
  • chlorocarbonylpolyfluoroalkylsulfenate esters
  • chlorocarbonylhexafluoroisopropylidenimino sulfenate
  • 5-tri-fluoromethyl-2-oxo-1,3,4-oxathiazole
  • commercially important N,N-dialkylcarbamoyl chlorides

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

143.6 °F - closed cup

Flash Point(C)

62 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Keith G McMillan et al.
Carbohydrate research, 341(1), 41-48 (2005-11-03)
5-(1,2,3,4-tetra-O-acetyl-alpha-D-xylopyranos-5S-C-yl)-1,3,4-oxathiazol-2-one (8) has been prepared from glucuronamide in two steps and 73% overall yield by conversion to the tetra-O-acetyl derivative 7 followed by reaction with chlorocarbonylsulfenyl chloride. 5-(2,3,4-Tri-O-acetyl-beta-D-xylopyranosyl)-1,3,4-oxathiazol-2-one (12) was synthesised from D-xylose by a four-step sequence involving conversion to
An improved one-pot cost-effective synthesis of N, N-disubstituted carbamoyl halides and derivatives.
Adeppa K, et al.
Canadian Journal of Chemistry, 88(!2), 1277-1280 (2010)
Some fluorinated heterocyclic and acyclic derivatives of chlorocarbonylsulfenyl chloride.
John EO and Jean'ne MS.
Journal of Fluorine Chemistry, 36(4), 429-438 (1987)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service