Skip to Content
Merck
All Photos(1)

Key Documents

152560

Sigma-Aldrich

BDPA complex with benzene(1:1), free radical

Synonym(s):

α,γ-Bisdiphenylene-β-phenylallyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H27
CAS Number:
Molecular Weight:
495.63
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:
NACRES:
NA.22

form

solid

Quality Level

mp

210 °C (dec.) (lit.)

SMILES string

C1(C([C]2C(C=CC=C3)=C3C4=C2C=CC=C4)=C5C(C=CC=C6)=C6C7=C5C=CC=C7)=CC=CC=C1.C8=CC=CC=C8

InChI

1S/C33H21.C6H6/c1-2-12-22(13-3-1)31(32-27-18-8-4-14-23(27)24-15-5-9-19-28(24)32)33-29-20-10-6-16-25(29)26-17-7-11-21-30(26)33;1-2-4-6-5-3-1/h1-21H;1-6H

InChI key

JLJWIVHYUCBSRC-UHFFFAOYSA-N

General description

BDPA complex with benzene (1:1) is a stable organic radical commonly employed in dynamic nuclear polarization NMR studies.

Application

BDPA complex with benzene (1:1), free radical is used to study the antiradical activities of some flavonols, flavones, flavanones, and flavan-3-ols.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electron spin relaxation and heterogeneity of the 1:1 ?,?-bisdiphenylene-?-phenylallyl (BDPA)/benzene complex
Mitchell DG, et al.
The Journal of Physical Chemistry B, 115(24), 7986-7990 (2011)
Kinetic study of flavonoid reactions with stable radicals
Butkovic V, et al.
Journal of Agricultural and Food Chemistry, 52(10), 2816-2820 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service