Merck
  • A novel RP-HPLC method for the determination of bharangin in Ghantu bharangi crude extracts.
A novel RP-HPLC method for the determination of bharangin in Ghantu bharangi crude extracts.
Pakistan journal of pharmaceutical sciences (2009-01-27)
Thadikamala Sathish, Pendyala Brahmaiah, Kancherla Sathya, Pullasi Bhojaraju, Nenavath Gopal Naik, Devarapalli Kezia, Reddy Shetty Prakasam
ABSTRACT
An accurate, simple, reproducible and sensitive RP-HPLC method for the determination of bharangin has been developed and validated. The separation of bharangin and 2-nitroaniline (internal standard) was achieved on Supelcosil LC-18 (3micro, 150 x 4.6 mm i.d.) column using UV detection at 388 nm. The mobile phase was consisting of methanol and 0.01 M KH(2)PO(4) buffer (pH 3.0, adjusted with ortho-phosphoric acid) (75:25, % v/v). The linear range of detection for bharangin was found to be 10-50 ng/ml. Intra-and inter-days assay relative standard deviations were less than 3.21. The method has been successfully applied to the determination of bharangin in various crude extracts. The method has been shown to be linear, reproducible, specific, and rugged.
Product Description
Supelco
SUPELCOSIL LC-18 Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm
Supelco
SUPELCOSIL LC-18 Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 3 mm
Supelco
SUPELCOSIL LC-18 Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm
Supelco
SUPELCOSIL LC-NH2-NP HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
SUPELCOSIL LC-18-S HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
SUPELCOSIL LC-18-S HPLC Column, 5 μm particle size, L × I.D. 25 cm × 2.1 mm
Supelco
SUPELCOSIL LC-18-S HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
SUPELCOSIL LC-NH2-NP Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm
Supelco
SUPELCOSIL LC-18 Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 2.1 mm