Merck
  • RNA binding protein Musashi-2 regulates PIWIL1 and TBX1 in mouse spermatogenesis.
RNA binding protein Musashi-2 regulates PIWIL1 and TBX1 in mouse spermatogenesis.
Journal of cellular physiology (2017-09-09)
Jessie M Sutherland, Alexander P Sobinoff, Barbara A Fraser, Kate A Redgrove, Nicole A Siddall, Peter Koopman, Gary R Hime, Eileen A McLaughlin
ABSTRACT
RNA-binding proteins (RBP) are important facilitators of post-transcriptional gene regulation. We have previously established that nuclear overexpression of the RBP Musashi-2 (MSI2) during male germ cell maturation is detrimental to sperm cell development and fertility. Herein we determine the genes and pathways impacted by the upregulation of Msi2. Microarray analysis and qPCR confirmed differential gene expression in factors fundamental to the cell cycle, cellular proliferation, and cell death. Similarly, comparative protein expression analysis via iTRAQ, immunoblot, and immunolocalization, identified differential expression and localization of important regulators of transcription, translation, RNA processing, and spermatogenesis. Specifically, the testis-expressed transcription factor, Tbx1, and the piRNA regulator of gamete development, Piwil1, were both found to be targeted for translational repression by MSI2. This study provides key evidence to support a fundamental role for MSI2 in post-transcriptional regulation during male gamete development.
Sigma-Aldrich
Anti-TM9SF2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Sigma-Aldrich
Magna RIP® RNA-Binding Protein Immunoprecipitation Kit, RNA Immunoprecipitation (RIP) Kit containing all necessary reagents to perform 12 individual RNA-binding protein immunoprecipitation (RIP) reactions using protein A/G magnetic beads.
Sigma-Aldrich
Anti-GAPDH antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution