Merck
Evaluation of a novel PXR-knockout in HepaRG
Pharmacology research & perspectives (2016-10-08)
Beth Williamson, Mathias Lorbeer, Michael D Mitchell, Timothy G Brayman, Robert J Riley
ABSTRACT
The nuclear pregnane X receptor (PXR) regulates the expression of genes involved in the metabolism, hepatobiliary disposition, and toxicity of drugs and endogenous compounds. PXR is a promiscuous nuclear hormone receptor (NHR) with significant ligand and DNA-binding crosstalk with the constitutive androstane receptor (CAR); hence, defining the precise role of PXR in gene regulation is challenging. Here, utilising a novel PXR-knockout (KO) HepaRG cell line, real-time PCR analysis was conducted to determine PXR involvement for a range of inducers. The selective PXR agonist rifampicin, a selective CAR activator, 6-(4-chlorophenyl)imidazo[2,1-b][1,3]thiazole-5-carbaldehyde O-(3,4-dichlorobenzyl)oxime (CITCO), and dual activators of CAR and PXR including phenobarbital (PB) were analyzed. HepaRG control cells (5F clone) were responsive to prototypical inducers of CYP2B6 and CYP3A4. No response was observed in the PXR-KO cells treated with rifampicin. Induction of CYP3A4 by PB, artemisinin, and phenytoin was also much reduced in PXR-KO cells, while the response to CITCO was maintained. This finding is in agreement with the abolition of functional PXR expression. The apparent EC
Sigma-Aldrich
HepaRG Thawing and Plating Medium Supplement with Antibiotics
Sigma-Aldrich
HepaRG Induction Medium Supplement with Antibiotics
Sigma-Aldrich
HepaRG Maintenance and Metabolism Medium Supplement with Antibiotics
Sigma-Aldrich
HepaRG Serum Free Induction Medium Supplement with Antibiotics
Sigma-Aldrich
Differentiated HepaRG Cells Cryopreserved
Sigma-Aldrich
HepaRG Serum Free Induction Medium Supplement
Sigma-Aldrich
HepaRG Thawing and Plating Medium Supplement
Sigma-Aldrich
HepaRG Maintenance and Metabolism Medium Supplement
Sigma-Aldrich
HepaRG Induction Medium Supplement