Merck
  • Energy stress-induced lncRNA FILNC1 represses c-Myc-mediated energy metabolism and inhibits renal tumor development.
Energy stress-induced lncRNA FILNC1 represses c-Myc-mediated energy metabolism and inhibits renal tumor development.
Nature communications (2017-10-06)
Zhen-Dong Xiao, Leng Han, Hyemin Lee, Li Zhuang, Yilei Zhang, Joelle Baddour, Deepak Nagrath, Christopher G Wood, Jian Gu, Xifeng Wu, Han Liang, Boyi Gan
ABSTRACT
The roles of long non-coding RNAs in cancer metabolism remain largely unexplored. Here we identify FILNC1 (FoxO-induced long non-coding RNA 1) as an energy stress-induced long non-coding RNA by FoxO transcription factors. FILNC1 deficiency in renal cancer cells alleviates energy stress-induced apoptosis and markedly promotes renal tumor development. We show that FILNC1 deficiency leads to enhanced glucose uptake and lactate production through upregulation of c-Myc. Upon energy stress, FILNC1 interacts with AUF1, a c-Myc mRNA-binding protein, and sequesters AUF1 from binding c-Myc mRNA, leading to downregulation of c-Myc protein. FILNC1 is specifically expressed in kidney, and is downregulated in renal cell carcinoma; also, its low expression correlates with poor clinical outcomes in renal cell carcinoma. Together, our study not only identifies FILNC1 as a negative regulator of renal cancer with potential clinical value, but also reveals a regulatory mechanism by long non-coding RNAs to control energy metabolism and tumor development.FoxO are commonly down-regulated transcription factors and tumor suppressors in renal cell cancer (RCC). Here, the authors show that upon energy stress FoxOs induce the expression of the long non-coding RNA FILNC1, which inhibits survival of RCC by downregulating c-Myc and c-Myc-dependent metabolic rewiring.
Sigma-Aldrich
AICAR, ≥98% (HPLC), powder
Sigma-Aldrich
Anti-Vinculin antibody, Mouse monoclonal, clone hVIN-1, purified from hybridoma cell culture
Sigma-Aldrich
RIPAb+ AUF1 - RIP Validated Antibody and Primer Set, from rabbit
Sigma-Aldrich
2-Deoxy-2-[(7-nitro-2,1,3-benzoxadiazol-4-yl)amino]-D-glucose, ≥97% (HPLC)
Sigma-Aldrich
Magna RIP® RNA-Binding Protein Immunoprecipitation Kit, RNA Immunoprecipitation (RIP) Kit containing all necessary reagents to perform 12 individual RNA-binding protein immunoprecipitation (RIP) reactions using protein A/G magnetic beads.