Merck
  • Intrinsic membrane properties and inhibitory synaptic input of kenyon cells as mechanisms for sparse coding?
Intrinsic membrane properties and inhibitory synaptic input of kenyon cells as mechanisms for sparse coding?
Journal of neurophysiology (2009-06-26)
Heike Demmer, Peter Kloppenburg
ABSTRACT
The insect mushroom bodies (MBs) are multimodal signal processing centers and are essential for olfactory learning. Electrophysiological recordings from the MBs' principal component neurons, the Kenyon cells (KCs), showed a sparse representation of olfactory signals. It has been proposed that the intrinsic and synaptic properties of the KC circuitry combine to reduce the firing of action potentials and to generate relatively brief windows for synaptic integration in the KCs, thus causing them to operate as coincidence detectors. To better understand the ionic mechanisms that mediate the KC intrinsic firing properties, we used whole cell patch-clamp recordings from KCs in the adult, intact brain of Periplaneta americana to analyze voltage- and/or Ca2+-dependent inward (ICa, INa) and outward currents [IA, IK(V), IK,ST, IO(Ca)]. In general the currents had properties similar to those of currents in other insect neurons. Certain functional parameters of ICa and IO(Ca), however, had unusually high values, allowing them to assist sparse coding. ICa had a low-activation threshold and a very high current density compared with those of ICa in other insect neurons. Together these parameters make ICa suitable for boosting and sharpening the excitatory postsynaptic potentials as reported in previous studies. IO(Ca) also had a large current density and a very depolarized activation threshold. In combination, the large ICa and IO(Ca) are likely to mediate the strong spike frequency adaptation. These intrinsic properties of the KCs are likely to be supported by their tonic, inhibitory synaptic input, which was revealed by specific GABA antagonists and which contributes significantly to the hyperpolarized membrane potential at rest.
Sigma-Aldrich
4-Aminopyridine, 98%
Sigma-Aldrich
Papain from papaya latex, lyophilized powder, ≥10 units/mg protein
Sigma-Aldrich
Hyaluronidase from bovine testes, Type I-S, lyophilized powder, 400-1000 units/mg solid
Sigma-Aldrich
(±)-Citronellal, natural, ≥85%, FCC, FG
Sigma-Aldrich
Tetraethylammonium chloride, ≥98% (titration)