Merck
  • Determination of thymopentin in beagle dog blood by liquid chromatography with tandem mass spectrometry and its application to a preclinical pharmacokinetic study.
Determination of thymopentin in beagle dog blood by liquid chromatography with tandem mass spectrometry and its application to a preclinical pharmacokinetic study.
Journal of separation science (2015-01-30)
Meiyun Shi, Yan Yang, Xiaotong Zhou, Lanlan Cai, Chunxue Fang, Can Wang, Heping Sun, Yantong Sun, Yin Gao, Jingkai Gu, J Paul Fawcett
ABSTRACT
The pentapeptide thymopentin (Arg-Lys-Asp-Val-Tyr, RKDVY) corresponds to amino acids 32-36 of the 49 amino acid immunomodulatory polypeptide, thymopoietin, whose biological activity is partially reproduced. Thymopentin is widely used in the clinic and represents a promising target for drug design but bioanalytical and pharmacokinetic data are limited due to its enzymatic instability. This paper reports a rapid and sensitive method based on liquid chromatography with tandem mass spectrometry for the determination of thymopentin in beagle dog blood. To inactivate peptidases and stabilize thymopentin, acetonitrile was added to blood samples immediately after collection followed by addition of stable isotope-labeled thymopentin as internal standard and washing with dichloromethane. Chromatography was carried out on an Ascentis Express Peptide ES-C18 column using gradient elution with methanol and aqueous 0.1% formic acid at a flow rate of 0.6 mL/min. Positive electrospray ionization mass spectrometry with selected reaction monitoring achieved linearity in the range of 1.5-800 ng/mL with good accuracy/precision and minimal matrix effects. The method was successfully applied to a pharmacokinetic study in beagle dogs after intravenous administration of 0.2 mg/kg thymopentin.
Product Description
Supelco
Ascentis® Express C18, 2.7 Micron Capillary HPLC Column, 2.7 μm particle size, L × I.D. 5 cm × 300 μm
Supelco
Ascentis® Express Peptide 160 Å ES-C18 (2.7 μm) HPLC Column, L × I.D. 3 cm × 4.6 mm, pk of 1 ea
Supelco
Ascentis® Express Peptide ES-C18, 2.7 μm Capillary HPLC Column, 2.7 μm particle size, L × I.D. 5 cm × 300 μm
Supelco
Ascentis® Express C18 (2.7 μm) HPLC Column, L × I.D. 5 cm × 1 mm, pk of 1 ea
Supelco
Ascentis® Express C18 (2.7 μm) HPLC Column, L × I.D. 7.5 cm × 2.1 mm, pk of 1 ea
Supelco
Ascentis® Express Peptide ES-C18, 2.7 μm Capillary HPLC Column, 2.7 μm particle size, L × I.D. 15 cm × 1 mm
Supelco
Ascentis® Express Peptide 160 Å ES-C18 (2.7 μm) HPLC Column, L × I.D. 15 cm × 3 mm, pk of 1 ea
Supelco
Ascentis® Express Peptide 160 Å ES-C18 (2.7 μm) HPLC Column, L × I.D. 15 cm × 4.6 mm, pk of 1 ea
Supelco
Ascentis® Express Peptide ES-C18, 2.7 μm Capillary HPLC Column, 2.7 μm particle size, L × I.D. 15 cm × 300 μm
Supelco
Ascentis® Express Peptide ES-C18, 2.7 μm Capillary HPLC Column, 2.7 μm particle size, L × I.D. 5 cm × 500 μm
Supelco
Ascentis® Express Peptide 160 Å ES-C18 (2.7 μm) HPLC Column, L × I.D. 7.5 cm × 4.6 mm, pk of 1 ea
Supelco
Ascentis® Express Peptide ES-C18, 2.7 μm Guard Cartridge, 2.7 μm particle size, L × I.D. 5 mm × 4.6 mm, pkg of 3 ea
Supelco
Ascentis® Express C18 (2.7 μm) HPLC Column, L × I.D. 15 cm × 4.6 mm, pk of 1 ea
Supelco
Ascentis® Express Peptide 160 Å ES-C18 (2.7 μm) HPLC Column, L × I.D. 5 cm × 4.6 mm, pk of 1 ea
Supelco
Ascentis® Express Peptide ES-C18, 2.7 μm Guard Cartridge, 2.7 μm particle size, L × I.D. 5 mm × 2.1 mm, pkg of 3 ea
Supelco
Ascentis® Express C18 (2.7 μm) HPLC Column, L × I.D. 10 cm × 3 mm, pk of
Supelco
Ascentis® Express Peptide 160 Å ES-C18 (2.7 μm) HPLC Column, L × I.D. 10 cm × 2.1 mm, pk of 1 ea
Supelco
Ascentis® Express Peptide 160 Å ES-C18 (2.7 μm) HPLC Column, L × I.D. 5 cm × 3 mm, pk of 1 ea
Supelco
Ascentis® Express Peptide 160 Å ES-C18 (2.7 μm) HPLC Column, L × I.D. 3 cm × 2.1 mm, pk of 1 ea
Supelco
Ascentis® Express Peptide 160 Å ES-C18 (2.7 μm) HPLC Column, L × I.D. 10 cm × 3 mm, pk of 1 ea
Supelco
Ascentis® Express C18 (2.7 μm) HPLC Column, L × I.D. 3 cm × 3 mm, pk of 1 ea
Supelco
Ascentis® Express C18 (2.7 μm) HPLC Column, L × I.D. 2 cm × 2.1 mm, pk of 1 ea
Supelco
Ascentis® Express C18 (2.7 μm) HPLC Column, L × I.D. 10 cm × 4.6 mm, pk of 1 ea
Supelco
Ascentis® Express C18 (2.7 μm) HPLC Column, L × I.D. 15 cm × 3 mm, pk of 1 ea
Supelco
Ascentis® Express C18 (2.7 μm) HPLC Column, L × I.D. 7.5 cm × 3 mm, pk of
Supelco
Ascentis® Express C18 (2.7 μm) HPLC Column, L × I.D. 5 cm × 4.6 mm, pk of 1 ea
Supelco
Ascentis® Express C18 (2.7 μm) HPLC Column, L × I.D. 5 cm × 2.1 mm, pk of 1 ea
Supelco
Ascentis® Express Peptide ES-C18, 2.7 μm Capillary HPLC Column, 2.7 μm particle size, L × I.D. 5 cm × 200 μm
Supelco
Ascentis® Express Peptide 160 Å ES-C18 (2.7 μm) HPLC Column, L × I.D. 10 cm × 4.6 mm, pk of 1 ea
Supelco
Ascentis® Express C18 (2.7 μm) HPLC Column, L × I.D. 3 cm × 2.1 mm, pk of 1 ea