Merck
  • Chloroquine triggers Epstein-Barr virus replication through phosphorylation of KAP1/TRIM28 in Burkitt lymphoma cells.
Chloroquine triggers Epstein-Barr virus replication through phosphorylation of KAP1/TRIM28 in Burkitt lymphoma cells.
PLoS pathogens (2017-03-02)
Xiaofan Li, Eric M Burton, Sumita Bhaduri-McIntosh
ABSTRACT
Trials to reintroduce chloroquine into regions of Africa where P. falciparum has regained susceptibility to chloroquine are underway. However, there are long-standing concerns about whether chloroquine increases lytic-replication of Epstein-Barr virus (EBV), thereby contributing to the development of endemic Burkitt lymphoma. We report that chloroquine indeed drives EBV replication by linking the DNA repair machinery to chromatin remodeling-mediated transcriptional repression. Specifically, chloroquine utilizes ataxia telangiectasia mutated (ATM) to phosphorylate the universal transcriptional corepressor Krüppel-associated Box-associated protein 1/tripartite motif-containing protein 28 (KAP1/TRIM28) at serine 824 -a mechanism that typically facilitates repair of double-strand breaks in heterochromatin, to instead activate EBV. Notably, activation of ATM occurs in the absence of detectable DNA damage. These findings i) clarify chloroquine's effect on EBV replication, ii) should energize field investigations into the connection between chloroquine and endemic Burkitt lymphoma and iii) provide a unique context in which ATM modifies KAP1 to regulate persistence of a herpesvirus in humans.
Sigma-Aldrich
2-Phenylindole, technical grade, 95%
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Duolink® In Situ Wash Buffers, Fluorescence
Sigma-Aldrich
Duolink® In Situ Detection Reagents Green
Sigma-Aldrich
KU-55933, ≥98% (HPLC)
Sigma-Aldrich
Sodium butyrate, 98%
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat MINUS, Affinity purified Donkey anti-Goat IgG (H+L)
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit PLUS, Affinity purified Donkey anti-Rabbit IgG (H+L)
Sigma-Aldrich
Goat Anti-Rabbit IgG Antibody, (H+L) HRP conjugate, 1 mg/mL, Chemicon®