Merck
  • Woodchuck hepatitis virus posttranscriptional regulatory element enhances expression of transgenes delivered by retroviral vectors.
Woodchuck hepatitis virus posttranscriptional regulatory element enhances expression of transgenes delivered by retroviral vectors.
Journal of virology (1999-03-12)
R Zufferey, J E Donello, D Trono, T J Hope
ABSTRACT
The expression of genes delivered by retroviral vectors is often inefficient, a potential obstacle for their widespread use in human gene therapy. Here, we explored the possibility that the posttranscriptional regulatory element of woodchuck hepatitis virus (WPRE) might help resolve this problem. Insertion of the WPRE in the 3' untranslated region of coding sequences carried by either oncoretroviral or lentiviral vectors substantially increased their levels of expression in a transgene-, promoter- and vector-independent manner. The WPRE thus increased either luciferase or green fluorescent protein production five- to eightfold, and effects of a comparable magnitude were observed with either the immediate-early cytomegalovirus or the herpesvirus thymidine kinase promoter and with both human immunodeficiency virus- and murine leukemia virus-based vectors. The WPRE exerted this influence only when placed in the sense orientation, consistent with its predicted posttranscriptional mechanism of action. These results demonstrate that the WPRE significantly improves the performance of retroviral vectors and emphasize that posttranscriptional regulation of gene expression should be taken into account in the design of gene delivery systems.
Sigma-Aldrich
MISSION® TRC2 pLKO.5-puro Empty Vector Control Plasmid DNA, Contains no shRNA insert
Sigma-Aldrich
MISSION® TRC2 pLKO.5-puro TurboGFP shRNA Control Plasmid DNA, shRNA sequence targeting tGFP
Sigma-Aldrich
MISSION® TRC2 pLKO.5-puro Non-Mammalian shRNA Control Plasmid DNA, Targets no known mammalian genes
Sigma-Aldrich
MISSION® TRC2 pLKO.5-puro Non-Mammalian shRNA Control Transduction Particles, Targets no known mammalian genes
Sigma-Aldrich
MISSION® TRC2 pLKO.5-puro-CMV-TurboGFP Positive Control Transduction Particles, Green fluorescent protein marker to monitor transduction efficiency
Sigma-Aldrich
MISSION® TRC2 pLKO.5-puro Empty Vector Control Transduction Particles puro, Contains no shRNA insert
Sigma-Aldrich
MISSION® TRC2 pLKO.5-puro-CMV-TurboGFP Positive Control Plasmid DNA, Green fluorescent protein marker to monitor transduction efficiency
Sigma-Aldrich
MISSION® TRC2 pLKO.5-puro TurboGFP shRNA Control Transduction Particles, shRNA sequence targeting tGFP