Merck
  • Phosphorylation of murine double minute-2 on Ser166 is downstream of VEGF-A in exercised skeletal muscle and regulates primary endothelial cell migration and FoxO gene expression.
Phosphorylation of murine double minute-2 on Ser166 is downstream of VEGF-A in exercised skeletal muscle and regulates primary endothelial cell migration and FoxO gene expression.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2015-11-19)
Julian Aiken, Emilie Roudier, Joseph Ciccone, Genevieve Drouin, Anna Stromberg, Jovana Vojnovic, I Mark Olfert, Tara Haas, Thomas Gustafsson, Guillaume Grenier, Olivier Birot
ABSTRACT
We demonstrated in a previous study that murine double minute (Mdm)-2 is essential for exercise-induced skeletal muscle angiogenesis. In the current study, we investigated the mechanisms that regulate Mdm2 activity in response to acute exercise and identified VEGF-A as a key stimulator of Mdm2 phosphorylation on Ser(166) (p-Ser166-Mdm2). VEGF-A and p-Ser166-Mdm2 protein levels were measured in human and rodent muscle biopsy specimens after 1 bout of exercise. VEGF-A-dependent Mdm2 phosphorylation was demonstrated in vivo in mice harboring myofiber-specific deletion of VEGF-A (mVEGF(-/-)) and in vitro in primary human and rodent endothelial cells (ECs). Exercise increased VEGF-A and p-Ser166-Mdm2 protein levels respectively by 157 and 68% in human muscle vs. pre-exercise levels. Similar results were observed in exercised rodent muscles compared to sedentary controls; however, exercise-induced Mdm2 phosphorylation was significantly attenuated in mVEGF(-/-) mice. Recombinant VEGF-A elevated p-Ser166-Mdm2 by 50-125% and stimulated migration by 33% in ECs when compared to untreated cells, whereas the Mdm2 antagonist Nutlin-3a abrogated VEGF-driven EC migration. Finally, overexpression of a Ser166-Mdm2 phosphorylation mimetic increased EC migration, increased Mdm2 to FoxO1 binding (+55%), and decreased FoxO1-dependent gene expression compared with ECs overexpressing WT-Mdm2. Our results suggest that VEGF-mediated Mdm2 phosphorylation on Ser(166) is a novel proangiogenic pathway within the skeletal muscle.
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
Giemsa Stain, Modified
Sigma-Aldrich
Anti-MDM2 (Ab-3) Mouse mAb (4B11), liquid, clone 4B11, Calbiochem®
Sigma-Aldrich
Nutlin-3a, ≥98% (HPLC)