Merck
  • Dye labelled monoclonal antibody assay for detection of Toxic Shock Syndrome Toxin -1 from Staphylococcus aureus.
Dye labelled monoclonal antibody assay for detection of Toxic Shock Syndrome Toxin -1 from Staphylococcus aureus.
Iranian journal of microbiology (2012-04-25)
Khojasteh V Javid, Ha Foster
ABSTRACT
The aim of study was to develop a rapid assay, dye labelled monoclonal antibody assay (DLMAA), using non-radioactive organic synthetic dyes for identification of Toxic Shock Syndrome Toxin-1 (TSST-1) producing strains of Staphylococcus aureus. The assay protocol required only two simple steps; addition of TSST-1 antigen to a nitrocellulose membrane and then adding a colloidal dye labelled antibody (D/A) suspension detection reagent. The sensitivity and specificity of the assay was determined relative to positive and negative strains compared to an ELISA assay. Overall 100% agreement was found between both assays. The sensitivity for detection of TSST-1 was 30 ng. The DLMAA did not require handling and disposal of radioactive materials. It is a rapid qualitative technique for detection of TSST-1 toxin at room temperature within a short time.
Sigma-Aldrich
Anti-Human IgG (γ-chain specific)−Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, tablet, 1 mg substrate per tablet