Merck
  • Identification of the novel activity-driven interaction between synaptotagmin 1 and presenilin 1 links calcium, synapse, and amyloid beta.
Identification of the novel activity-driven interaction between synaptotagmin 1 and presenilin 1 links calcium, synapse, and amyloid beta.
BMC biology (2016-04-03)
Akira Kuzuya, Katarzyna M Zoltowska, Kathryn L Post, Muriel Arimon, Xuejing Li, Sarah Svirsky, Masato Maesako, Alona Muzikansky, Vivek Gautam, Dora Kovacs, Bradley T Hyman, Oksana Berezovska
ABSTRACT
Synaptic loss strongly correlates with memory deterioration. Local accumulation of amyloid β (Aβ) peptide, and neurotoxic Aβ42 in particular, due to abnormal neuronal activity may underlie synaptic dysfunction, neurodegeneration, and memory impairments. To gain an insight into molecular events underlying neuronal activity-regulated Aβ production at the synapse, we explored functional outcomes of the newly discovered calcium-dependent interaction between Alzheimer's disease-associated presenilin 1 (PS1)/γ-secretase and synaptic vesicle proteins. Mass spectrometry screen of mouse brain lysates identified synaptotagmin 1 (Syt1) as a novel synapse-specific PS1-binding partner that shows Ca(2+)-dependent PS1 binding profiles in vitro and in vivo. We found that Aβ level, and more critically, conformation of the PS1 and the Aβ42/40 ratio, are affected by Syt1 overexpression or knockdown, indicating that Syt1 and its interaction with PS1 might regulate Aβ production at the synapse. Moreover, β-secretase 1 (BACE1) stability, β- and γ-secretase activity, as well as intracellular compartmentalization of PS1 and BACE1, but not of amyloid precursor protein (APP), nicastrin (Nct), presenilin enhancer 2 (Pen-2), or synaptophysin (Syp) were altered in the absence of Syt1, suggesting a selective effect of Syt1 on PS1 and BACE1 trafficking. Our findings identify Syt1 as a novel Ca(2+)-sensitive PS1 modulator that could regulate synaptic Aβ, opening avenues for novel and selective synapse targeting therapeutic strategies.
Sigma-Aldrich
Anti-Pen-2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Amyloid Precursor Protein, C-Terminal antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Nicastrin (ab2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Presenilin-1 (S182) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Presenilin-1 Antibody, NT, clone hPS1-NT, culture supernatant, clone hPS1-NT, Chemicon®
Sigma-Aldrich
Anti-APP A4 Antibody, a.a. 66-81 of APP {NT}, clone 22C11, clone 22C11, Chemicon®, from mouse
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-Synaptotagmin I Antibody, Chemicon®, from mouse