Merck
  • Quantification of Salivary Arecoline, Arecaidine and N-Methylnipecotic Acid Levels in Volunteers by Liquid Chromatography-Tandem Mass Spectrometry.
Quantification of Salivary Arecoline, Arecaidine and N-Methylnipecotic Acid Levels in Volunteers by Liquid Chromatography-Tandem Mass Spectrometry.
Journal of analytical toxicology (2015-08-02)
Hei Hwa Lee, Li Yao Chen, Hui Liang Wang, Bai Hsiun Chen
ABSTRACT
Relatively little is known about the metabolism of areca nut in human saliva. We here describe the simultaneous quantification of areca nut metabolites: arecoline, arecaidine and N-methylnipecotic acid in saliva samples after chewing one 5 g areca nut by using liquid chromatography-tandem mass spectrometry (LC-MS-MS). Time courses of salivary areca nut metabolites in five adult male areca nut chewer volunteers were investigated. The limits of quantification were all 1.25 ng/mL for arecoline, arecaidine and N-methylnipecotic acid. Intra- and interday imprecisions were <4.2 and 13.6%, respectively. The within-day accuracy ranged from 82.2 to 116.7%, and the between-day accuracy ranged from 78.3 to 115.6%. Through areca nut chewing time course study, we found that salivary arecoline, arecaidine and N-methylnipecotic acid concentrations varied greatly over time between experiment individuals. Our findings suggest that arecoline might be metabolized slightly to arecaidine at 30 min after areca nut chewing and arecoline might be metabolized slightly to N-methylnipecotic acid at 25 min after areca nut chewing in the mouth. We first provide simultaneous quantification of human salivary arecoline, arecaidine and N-methylnipecotic acid levels using LC-MS-MS. This method may facilitate future research design in the pathogenic effects of areca nut exposure.
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
1-methylpiperidine-3-carboxylic acid, AldrichCPR
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, for chromatography