Merck
  • Loading and removal of PAHs in a wastewater treatment plant in a separated sewer system.
Loading and removal of PAHs in a wastewater treatment plant in a separated sewer system.
Water research (2015-06-01)
Noriatsu Ozaki, Yoshihiro Takamura, Keisuke Kojima, Tomonori Kindaichi
ABSTRACT
The loading and removal of polycyclic aromatic hydrocarbons (PAHs) were measured and estimated in a wastewater treatment plant in a separated sewer system in a suburban area of Japan. The influent 16 PAHs concentration was 219 ± 210 ng L(-1), whereas the effluent concentration was 43.5 ± 42.5 ng L(-1) (mean ± sd). No clear diurnal or weekly fluctuation was observed. However, evaluation of long-term changes revealed PAH fluctuations continuing for more than 1 week. Half of the PAHs (63%) were biologically or chemically transformed, or vaporized in the treatment plant, while the remainder were discharged with effluent (28%) and excess sludge (9%). Measurement of the per capita loading of the treatment plant revealed values of 142 ± 53 and 28 ± 11 μg person(-1)day(-1) (mean ± 95% confidence interval) for influent and effluent, respectively. Isomer ratio analysis revealed that the PAHs originated from a mixture of petroleum, petroleum combustion, and burning of biomass residues.
Sigma-Aldrich
Fluorene, 98%
Sigma-Aldrich
Pyrene, puriss. p.a., for fluorescence, ≥99.0% (GC)
Sigma-Aldrich
Pyrene, 98%
Sigma-Aldrich
Benzo[ghi]perylene, 98%
Sigma-Aldrich
Anthracene, suitable for scintillation, ≥99.0% (GC)
Sigma-Aldrich
Acenaphthene-d10, 99 atom % D
Sigma-Aldrich
Anthracene, ReagentPlus®, 99%
Sigma-Aldrich
Anthracene, reagent grade, 97%
Sigma-Aldrich
Benzo[k]fluoranthene, for fluorescence, ≥99%
Sigma-Aldrich
2-Fluorobiphenyl, 96%
Sigma-Aldrich
Acenaphthylene, 75%
Sigma-Aldrich
Pyrene, sublimed grade, 99%
Sigma-Aldrich
Benzo[b]fluoranthene, 98%
Sigma-Aldrich
Acenaphthylene, 99%
Sigma-Aldrich
Chrysene-d12, 98 atom % D
Sigma-Aldrich
Anthracene, sublimed grade, ≥99%
Sigma-Aldrich
Perylene-d12, 98 atom % D
Sigma-Aldrich
Benzo[a]pyrene, ≥96% (HPLC)
Sigma-Aldrich
Benz[a]anthracene, 99%
Sigma-Aldrich
p-Terphenyl-d14, 98 atom % D, 98% (CP)
Sigma-Aldrich
Phenanthrene-d10, 98 atom % D, 98% (CP)
Sigma-Aldrich
Fluoranthene, 98%
Sigma-Aldrich
Benzo[e]pyrene, 98%
Sigma-Aldrich
Acenaphthene, 99%
Sigma-Aldrich
Phenanthrene, 98%
Sigma-Aldrich
Phenanthrene, sublimed grade, ≥99.5%