Merck
  • Water-soluble nanoconjugates of quantum dot-chitosan-antibody for in vitro detection of cancer cells based on "enzyme-free" fluoroimmunoassay.
Water-soluble nanoconjugates of quantum dot-chitosan-antibody for in vitro detection of cancer cells based on "enzyme-free" fluoroimmunoassay.
Materials science & engineering. C, Materials for biological applications (2015-05-09)
Herman S Mansur, Alexandra A P Mansur, Amanda Soriano-Araújo, Zélia I P Lobato, Sandhra M de Carvalho, Maria de Fatima Leite
ABSTRACT
Cancer remains one of the world's most devastating diseases with millions of fatalities and new cases every year. In this work, we attempted to develop a facile "enzyme-free" fluoroimmunoassay based on the novel nanoconjugates composed of CdS quantum dots (QDs) as the fluorescent inorganic core and an antibody-modified polysaccharide as the organic shell, modeling their possible application for the in vitro diagnosis of non-Hodgkin lymphoma (NHL) cancer. Chitosan was conjugated with an anti-CD20 polyclonal antibody (pAbCD20) by the formation of covalent amide bonds. In the sequence, these chitosan-antibody conjugates were utilized as direct ligands for the surface biofunctionalization of CdS QDs (CdS/chitosan-pAbCD20) using a single-step colloidal process in aqueous medium at room temperature. The most relevant physico-chemical properties of these nanoconjugates were assessed by morphological and spectroscopic techniques. The results indicated that CdS nanocrystals were produced with an average diameter of 2.5 nm and with cubic zinc blende crystalline nanostructure. The CdS-immunoconjugates (CdS/chitosan-pAbCD20) presented colloidal hydrodynamic diameter (HD) of 15.0 ± 1.2n m. In addition, the results evidenced that the "enzyme-free" QD-linked immunosorbent assay (QLISA) was effective for the in vitro detection against the antigen CD20 (aCD20) based on fluorescent behavior of the CdS nanoconjugates. Moreover, the CdS-immunoconjugates were successfully used for fluorescence bioimaging of NHL cancer cells. Finally, the cell viability results using different cell cultures based on LDH, MTT and Resazurin bio-assays have demonstrated no cytotoxicity of the new CdS-chitosan bioconjugates relative to the standard controls. Thus, CdS conjugates may offer a promising platform for the future development of in vitro and in vivo applications for the detection and diagnosis of NHL cancer cells.
Sigma-Aldrich
Acetic acid solution, 1 M, 1 N
Sigma-Aldrich
Acetic acid, ≥99.7%, suitable for amino acid analysis
Sigma-Aldrich
Sodium hydroxide solution, 0.01 M
Sigma-Aldrich
Sodium hydroxide solution, 0.1 M
Sigma-Aldrich
Sodium hydroxide solution, 7 M
Sigma-Aldrich
Sodium hydroxide solution, 4 M
Sigma-Aldrich
Acetic acid, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetic acid, 99.5-100.0%
Sigma-Aldrich
Sodium hydroxide solution, 0.05 M
Sigma-Aldrich
Sodium hydroxide, JIS special grade, ≥96.0%
Sigma-Aldrich
Sodium hydroxide solution, 6 M
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Cadmium perchlorate hydrate, 99.999% trace metals basis
Sigma-Aldrich
Rhodamine 6G, Dye content 99 %
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Streptomycin sulfate salt, powder
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Rhodamine 6G, Dye content ~95 %
Sigma-Aldrich
Cadmium perchlorate hydrate
Sigma-Aldrich
Sodium sulfide hydrate, ≥60%, scales
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium sulfide
Sigma-Aldrich
Rhodamine 6G, suitable for fluorescence, BioReagent
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets