Merck
  • Living Composites of Bacteria and Polymers as Biomimetic Films for Metal Sequestration and Bioremediation.
Living Composites of Bacteria and Polymers as Biomimetic Films for Metal Sequestration and Bioremediation.
Macromolecular bioscience (2015-04-08)
Christian Knierim, Michaela Enzeroth, Patrick Kaiser, Christian Dams, David Nette, Andreas Seubert, Andreas Klingl, Charles L Greenblatt, Valérie Jérôme, Seema Agarwal, Ruth Freitag, Andreas Greiner
ABSTRACT
Herein, we report on composite materials of biologically active microorganisms placed in a synthetic polymer matrix. These so-called "living composites" were utilized for gold sequestration (Micrococcus luteus) and bioremediation of nitrite (Nitrobacter winogradskyi) to demonstrate functionality. For the preparation of the living composites the bacteria were first encased in a water-soluble polymer fiber (poly(vinyl alcohol), PVA) followed by coating the fibers with a shell of hydrophobic poly(p-xylylene) (PPX) by chemical vapor deposition (CVD). The combination of bacteria with polymer materials assured the stability and biologically activity of the bacteria in an aqueous environment for several weeks.
Sigma-Aldrich
Acetone, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetone, ≥99.5%, for residue analysis
Sigma-Aldrich
Acetone, for residue analysis, ≥99.5%
Sigma-Aldrich
Acetone, for residue analysis, JIS 5000
Sigma-Aldrich
Acetone, suitable for HPLC
Sigma-Aldrich
Acetone, JIS special grade, ≥99.5%
Sigma-Aldrich
Rhodamine 6G, suitable for fluorescence, BioReagent
Sigma-Aldrich
[2.2]Paracyclophane, 97%
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Rhodamine 6G, Dye content 99 %
Sigma-Aldrich
Rhodamine 6G, Dye content ~95 %
Sigma-Aldrich
Acetone, for chromatography, ≥99.8%