Merck
  • Investigation of polycyclic aromatic hydrocarbons (PAHs) and cyclic terpenoid biomarkers in the sediments of fishing harbors in Taiwan.
Investigation of polycyclic aromatic hydrocarbons (PAHs) and cyclic terpenoid biomarkers in the sediments of fishing harbors in Taiwan.
Marine pollution bulletin (2015-06-08)
Nien-Hsin Kao, Ming-Chien Su, Jheng-Rong Fan, Chih-Chun Yen
ABSTRACT
Three fishing harbors were investigated to study the polycyclic aromatic hydrocarbons in the sediments and trace possible anthropogenic sources by identification of cyclic terpenoid biomarkers. Seventeen terpanes, 10 steranes and 10 bicyclic sesquiterpanes in the marine diesel and the three kinds of lubricants that are mainly used by fishing boats were identified and quantified. Eighteen biomarker diagnostic ratios are suggested and the correlation coefficients among the lubricants and sediment samples have the R(2) value greater than 0.73. Analyzed 16 PAHs in the sediment shows non-normal distributions and the Kruskal Wallis Test shows the significant differences (p value smaller than 0.05) with the greatest variability in benzo[g,h,i]perylene which more than 84% of the effective size (E.S.) is accounted. X-ray Photoelectron Spectroscopy (XPS) analysis was applied and the Kruskal Wallis Test shows a significant difference (p value smaller than 0.05) among certain atoms with the effective size greater than 60%.
Sigma-Aldrich
Acetone, for chromatography, ≥99.8%
Sigma-Aldrich
Acetone, for residue analysis, JIS 5000
Sigma-Aldrich
Acetone, suitable for HPLC
Sigma-Aldrich
Acetone, ≥99.5%, for residue analysis
Sigma-Aldrich
Acetone, for residue analysis, ≥99.5%
Sigma-Aldrich
Acetone, JIS special grade, ≥99.5%
Sigma-Aldrich
Acetone, SAJ first grade, ≥99.0%
Sigma-Aldrich
2,6-Dimethylnaphthalene, 99%
Sigma-Aldrich
2-Fluorobiphenyl, 96%
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
1,3-Dimethylnaphthalene, 96%
Sigma-Aldrich
1,6-Dimethylnaphthalene, 99%
Sigma-Aldrich
1,4-Dimethylnaphthalene, 95%
Sigma-Aldrich
Acenaphthene-d10, 99 atom % D
Sigma-Aldrich
1,2-Dimethylnaphthalene, 95%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Hexane, for residue analysis, JIS 5000
Sigma-Aldrich
Hexane, JIS 300, ≥96.0%, for residue analysis
Sigma-Aldrich
Hexane, SAJ first grade, ≥95.0%
Sigma-Aldrich
Hexane, JIS special grade, ≥96.0%
Sigma-Aldrich
Hexane, JIS 1000, ≥96.0%, for residue analysis
Sigma-Aldrich
2-Ethylnaphthalene, ≥99%
Sigma-Aldrich
2,7-Dimethylnaphthalene, 99%
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Hexane, ≥96.0%, suitable for residual phthalate analysis
Sigma-Aldrich
Hexane, suitable for HPLC