Merck
  • Young male mating success is associated with sperm number but not with male sex pheromone titres.
Young male mating success is associated with sperm number but not with male sex pheromone titres.
Frontiers in zoology (2015-11-12)
Tobias Kehl, Ian A N Dublon, Klaus Fischer
ABSTRACT
Intraspecific communication is of crucial importance throughout the animal kingdom and may involve a combination of visual, gustatory, olfactory and acoustic cues. Variation in male sex pheromone amount and composition may convey important information to female conspecifics, for instance on species identity or age. However, whether increased male pheromone titres are associated with fitness benefits for the female, thus indicating a role as an honest signal, is under debate. Against this background, we tested in the butterfly Bicyclus anynana (1) whether young males being successful or unsuccessful in gaining a mating differed in sex pheromone titres and (2) for associations between male pheromone titres and spermatophore mass, eupyrene sperm number, and a variety of female and offspring life-history traits. Successful and unsuccessful males did not differ in pheromone titres, however eupyrene sperm number was much higher in successful males. Pheromone titres were not associated with any fitness-related female or offspring trait measured in our study, though correlation analyses yielded evidence for trade-offs among specific traits. Patterns did not differ among control and olfaction-blocked females. Therefore, we suggest that in young B. anynana pheromone titres do not indicate male quality.
Sigma-Aldrich
trans-4-Tridecenyl acetate, AldrichCPR
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Hexane, suitable for HPLC
Sigma-Aldrich
Hexane, for residue analysis, JIS 5000
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Hexane, JIS special grade, ≥96.0%
Sigma-Aldrich
Hexane, SAJ first grade, ≥95.0%
Sigma-Aldrich
Hexane, JIS 300, ≥96.0%, for residue analysis
Sigma-Aldrich
Hexane, ≥96.0%, suitable for residual phthalate analysis
Sigma-Aldrich
Hexane, JIS 1000, ≥96.0%, for residue analysis
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%