Merck
  • Male obesity is associated with changed spermatozoa Cox4i1 mRNA level and altered seminal vesicle fluid composition in a mouse model.
Male obesity is associated with changed spermatozoa Cox4i1 mRNA level and altered seminal vesicle fluid composition in a mouse model.
Molecular human reproduction (2015-03-04)
Natalie K Binder, John R Sheedy, Natalie J Hannan, David K Gardner
ABSTRACT
The rate of obesity among men of reproductive age has tripled in the last three decades. Previously, we demonstrated that paternal obesity resulted in impaired preimplantation developmental kinetics, compromised post-compaction metabolism and decreased blastocyst cell number when embryos were generated in vivo. Subsequently, using in vitro fertilization we found embryos of obese males to have altered metabolism before compaction, reduced inner cell mass cell number and retarded fetal development--the difference between these two studies being the method of embryo generation and the presence or absence of seminal plasma, respectively. Here, we hypothesize that both sperm and seminal plasma are affected by obesity, compromising embryogenesis and pregnancy health in a cumulative manner. Epididymal sperm and seminal vesicle fluid were collected from normal and obese C57BL/6 mice. RNA and DNA were extracted from spermatozoa for qPCR and global methylation analysis, respectively. Proteomic (Luminex) and metabolomic (GC-MS) techniques were employed to analyse the composition of seminal vesicle fluid. Nuclear encoded cytochrome c oxidase subunit IV isoform 1 (Cox4i1) of the terminal enzyme in the mitochondrial respiratory chain demonstrated significantly increased RNA levels in the sperm of obese males (P< 0.05). Quantitative seminal plasma analysis identified significant changes in levels of the hormones insulin, leptin and estradiol between normal and obese males (P < 0.05). Further, the metabolite composition of seminal vesicle fluid was significantly affected by obesity. Consequently, this study has determined that obesity affects both sperm and seminal plasma composition. The interaction between sperm and seminal plasma warrants further analysis.
Sigma-Aldrich
Ethanol, 94.8-95.8%
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Sodium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
2-Hydroxybutyric acid sodium salt, 97%
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, ≥98.5% (titration)
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Sodium phosphate dibasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Ethanol, ≥99.5%, SAJ super special grade
Sigma-Aldrich
Citric acid, SAJ first grade, ≥99.5%
Sigma-Aldrich
Ethanol, JIS 300, ≥99.5%
Sigma-Aldrich
Ethanol, JIS 1000, ≥99.5%
Sigma-Aldrich
Ethanol, ≥99.5%
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for absorption spectrum analysis
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for HPLC
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for fluorescence
Sigma-Aldrich
Sodium phosphate dibasic, BioUltra, ≥99.5% (T)