Merck
RNF8 deficiency results in neurodegeneration in mice.
Neurobiology of aging (2015-08-11)
Siwei Ouyang, Yanfeng Song, Yingxia Tian, Yibin Chen, Xiaochun Yu, Degui Wang
ABSTRACT
The progressive loss of neurons causes neurodegenerative diseases. Because the accumulation of DNA breaks results in neuronal apoptosis, the lack of a variety of DNA damage repair-related proteins contributes to neurodegeneration. The ubiquitin ligase RNF8 plays an important role in DNA double-strand break repair via histone ubiquitination. However, the function of RNF8 in terminally differentiated neurons remains unknown. This study aimed to determine whether RNF8 is involved in the DNA damage response in neurons and contributes to neurodegeneration. Here, we present evidence suggesting that RNF8 deficiency results in DNA damage accumulation and neuronal apoptosis. RNF8(-/-) mice exhibit neuronal degeneration and reactive astrocytosis. Neurons from RNF8(-/-) mice appear to be more susceptible to X-ray-induced DNA damage. These changes were consistent with the behavioral performances of the RNF8-deficient mice, which included impaired performances in the open-field test and step-down avoidance task. Overall, these findings show that RNF8 is required for DNA damage repair in neurons. RNF8 deficiency is sufficient to cause neuronal pathology and cognitive decline, and the loss of RNF8 results in neuron degeneration.
Sigma-Aldrich
Anti-Histone H4 Antibody, Upstate®, from rabbit
Sigma-Aldrich
Goat Anti-Rabbit IgG Antibody, biotin-SP conjugate, Chemicon®, from goat
Sigma-Aldrich
Anti-Histone H2A Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein (GFAP) Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-ubiquityl-Histone H2A Antibody, clone E6C5, clone E6C5, Upstate®, from mouse
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Anti-Histone H2B Antibody, serum, Chemicon®