Merck
  • Effects of quaternization on the morphological stability and antibacterial activity of electrospun poly(DMAEMA-co-AMA) nanofibers.
Effects of quaternization on the morphological stability and antibacterial activity of electrospun poly(DMAEMA-co-AMA) nanofibers.
Colloids and surfaces. B, Biointerfaces (2015-06-22)
Jing-Wei Xu, Yao Wang, Yun-Feng Yang, Xiang-Yu Ye, Ke Yao, Jian Ji, Zhi-Kang Xu
ABSTRACT
Electrospun nanofibers with antibacterial activity are greatly promising for medical treatment and water purification. Herein we report antibacterial nanofibers electrospun from a series of poly(dimethylamino ethyl methacrylate-co-alkyl methacrylates) (poly(DMAEMA-co-AMA)) and to distinguish the effects of free and cross-linked cations derived from quanternization on the antibacterial activity. Poly(DMAEMA-co-AMA)s are simply synthesized by free radical polymerization from commercial monomers. DSC analysis indicates that they have Tg lower than room temperature and thus the electrospun nanofibers adhere to each other and evenly tend to form films, instead of keeping cylinderic shape. Benzyl chloride (BC) and p-xylylene dichloride (XDC) can quaternize DMAEMA units and to generate cations on the nanofiber surface. XPS analysis and colorimetric assay determine the quaternization degree and the surface accessible quaternary amines (N(+)), respectively. It is very promising that this quaternization endows the electrospun nanofibers with both stable morphology and antibacterial activity. The BC-quaternized fibers show better antibacterial behavior against Escherichia coli and Staphylococcus aureus than those of the XDC-quaternized/cross-linked ones, because cross-linking suppresses the chain mobility of cations. Our results confirm that antibacterial nanofibers can be facilely prepared and chain mobility of the formed cations is the necessary prerequisite for their antibacterial activity.
Sigma-Aldrich
Chloroform, suitable for HPLC
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer
Sigma-Aldrich
Cetyltrimethylammonium chloride solution, 25 wt. % in H2O
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
α,α′-Dichloro-p-xylene, 98%
Sigma-Aldrich
Chloroform, JIS 300, ≥99.0%
Sigma-Aldrich
Chloroform, SAJ super special grade, ≥99.0%
Sigma-Aldrich
Chloroform, SAJ first grade, ≥99.0%, contains 0.4-0.8% ethanol
Sigma-Aldrich
Chloroform, JIS special grade, ≥99.0%
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium chloride, ≥98.0% (NT)
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
1,3-Propylene sulfite, 99%
Sigma-Aldrich
Lauryl methacrylate, contains 500 ppm MEHQ as inhibitor, 96%
Sigma-Aldrich
Fluorescein sodium salt, used as fluorescent tracer
Sigma-Aldrich
Fluorescein sodium salt, BioReagent, suitable for fluorescence
Sigma-Aldrich
Fluorescein sodium salt, p.a.
Sigma-Aldrich
Butyl methacrylate, 99%, contains monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
Benzyl chloride, ReagentPlus®, 99%, contains ≤1% propylene oxide as stabilizer
Sigma-Aldrich
5-(N,N-Hexamethylene)amiloride
Sigma-Aldrich
Fluorescein sodium salt, JIS special grade