Merck
The mitochondrial carrier SLC25A10 regulates cancer cell growth.
Oncotarget (2015-03-24)
Xiaoshan Zhou, João A Paredes, Shuba Krishnan, Sophie Curbo, Anna Karlsson
ABSTRACT
Dysregulation of cell metabolism is critical for the growth properties of cancer cells. The purpose of this study was to understand the role of substrate transport across the mitochondrial membrane to sustain the metabolic shift and redox defense in cancer cells. Mitochondrial carrier SLC25A10 is up-regulated in a variety of tumors and is involved in regulating intracellular levels of reactive oxygen species. We show that knockdown of SLC25A10 in A549 cells changed the growth properties to a less malignant phenotype and casued increased glutamine dependency and sensitivity to oxidative stress. The metabolic alteration was linked to an energy metabolic shift from glycolysis to mitochondrial oxidative phosphorylation illustrated by increased expression of glutamate dehydrogenase, decreased expression of lactate dehydrogenase due to down-regulation of hypoxia inducible factor 1α. We identified effects on NADPH production linked to the growth changes observed in SLC25A10 knockdown cells, demonstrated by decreased NADPH production in cells deprived of glutamine. The contribution of SLC25A10 to reprogram cell metabolism and to regulate cell growth suggests SLC25A10 as a novel target for anti-cancer strategies.
Sigma-Aldrich
L-Glutamine
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Anti-SLC25A10 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Gemcitabine hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
SAFC
BIS-TRIS
SAFC
BIS-TRIS