Merck
  • CD24(hi)CD27⁺ and plasmablast-like regulatory B cells in human chronic graft-versus-host disease.
CD24(hi)CD27⁺ and plasmablast-like regulatory B cells in human chronic graft-versus-host disease.
Blood (2015-01-22)
Adèle de Masson, Jean-David Bouaziz, Hélène Le Buanec, Marie Robin, Alix O'Meara, Nathalie Parquet, Michel Rybojad, Estelle Hau, Jean-Benoît Monfort, Mylène Branchtein, David Michonneau, Valérie Dessirier, Flore Sicre de Fontbrune, Anne Bergeron, Raphaël Itzykson, Nathalie Dhédin, Djaouida Bengoufa, Régis Peffault de Latour, Aliénor Xhaard, Martine Bagot, Armand Bensussan, Gérard Socié
ABSTRACT
Interleukin 10 (IL-10)-producing B cells (regulatory B cells [Bregs]) regulate autoimmunity in mice and humans, and a regulatory role of IL-10-producing plasma cells has been described in mice. Dysfunction of B cells that maintain homeostasis may play a role in the pathogenesis of chronic graft-versus-host disease (cGVHD) after allogeneic stem cell transplantation. Here, we found a relation between decreased Breg frequencies and cGVHD severity. An impaired ability of B cells to produce IL-10, possibly linked to poor signal transducer and activator of transcription 3 and extracellular signal-regulated kinase phosphorylation, was found in patients with active cGVHD. IL-10 production was not confined to a single B-cell subset, but enriched in both the CD24(hi)CD27(+) and CD27(hi)CD38(hi) plasmablast B-cell compartments. In vitro plasmablast differentiation increased the frequency of IL-10-producing B cells. We confirmed that allogeneic transplant recipients had an impaired reconstitution of the memory B-cell pool. cGVHD patients had less CD24(hi)CD27(+) B cells and IL-10-producing CD24(hi)CD27(+) B cells. Patients with cGVHD had increased plasmablast frequencies but decreased IL-10-producing plasmablasts. These results suggest a role of CD24(hi)CD27(+) B-cell and plasmablast-derived IL-10 in the regulation of human cGVHD.
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Aphidicolin from Nigrospora sphaerica, ≥98% (HPLC), powder
Sigma-Aldrich
IL-21 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
IL-21 human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture