Merck
  • Effects of metal exposure on motor neuron development, neuromasts and the escape response of zebrafish embryos.
Effects of metal exposure on motor neuron development, neuromasts and the escape response of zebrafish embryos.
Neurotoxicology and teratology (2015-05-27)
Laura Sonnack, Sebastian Kampe, Elke Muth-Köhne, Lothar Erdinger, Nicole Henny, Henner Hollert, Christoph Schäfers, Martina Fenske
ABSTRACT
Low level metal contaminations are a prevalent issue with often unknown consequences for health and the environment. Effect-based, multifactorial test systems with zebrafish embryos to assess in particular developmental toxicity are beneficial but rarely used in this context. We therefore exposed wild-type embryos to the metals copper (CuSO4), cadmium (CdCl2) and cobalt (CoSO4) for 72 h to determine lethal as well as sublethal morphological effects. Motor neuron damage was investigated by immunofluorescence staining of primary motor neurons (PMNs) and secondary motor neurons (SMNs). In vivo stainings using the vital dye DASPEI were used to quantify neuromast development and damage. The consequences of metal toxicity were also assessed functionally, by testing fish behavior following tactile stimulation. The median effective concentration (EC50) values for morphological effects 72 h post fertilization (hpf) were 14.6 mg/L for cadmium and 0.018 mg/L for copper, whereas embryos exposed up to 45.8 mg/L cobalt showed no morphological effects. All three metals caused a concentration-dependent reduction in the numbers of normal PMNs and SMNs, and in the fluorescence intensity of neuromasts. The results for motor neuron damage and behavior were coincident for all three metals. Even the lowest metal concentrations (cadmium 2mg/L, copper 0.01 mg/L and cobalt 0.8 mg/L) resulted in neuromast damage. The results demonstrate that the neuromast cells were more sensitive to metal exposure than morphological traits or the response to tactile stimulation and motor neuron damage.
Sigma-Aldrich
Copper(II) sulfate, anhydrous, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Copper(II) sulfate, JIS first grade, ≥97.5%
Sigma-Aldrich
Neomycin trisulfate salt hydrate, powder
Sigma-Aldrich
Neomycin trisulfate salt hydrate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Neomycin trisulfate salt hydrate, meets USP testing specifications, powder
Sigma-Aldrich
Neomycin trisulfate salt hydrate, suitable for plant cell culture
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Ethanol, JIS first grade, 94.8-95.8%
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Sigma-Aldrich
trans-Platinum(II)diammine dichloride
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Sigma-Aldrich
Cadmium chloride, 99.99% trace metals basis
Sigma-Aldrich
Cadmium chloride, technical grade
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for absorption spectrum analysis
Sigma-Aldrich
cobalt(ii) sulfate heptahydrate, AldrichCPR
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for HPLC
Sigma-Aldrich
Ethanol, ≥99.5%
Sigma-Aldrich
Ethanol, ≥99.5%, SAJ super special grade
Sigma-Aldrich
Ethanol, JIS 300, ≥99.5%, for residue analysis
Sigma-Aldrich
Ethanol, JIS 1000, ≥99.5%, for residue analysis
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for fluorescence
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%