Merck
  • Potential market for novel tuberculosis diagnostics: worth the investment?
Potential market for novel tuberculosis diagnostics: worth the investment?
The Journal of infectious diseases (2015-03-15)
Sandra V Kik, Claudia M Denkinger, Carole Jefferson, Janet Ginnard, Madhukar Pai
ABSTRACT
The potential available market (PAM) for new diagnostics for tuberculosis that meet the specifications of the high-priority target product profiles (TPPs) is currently unknown. We estimated the PAM in 2020 in 4 high-burden countries (South Africa, Brazil, China, and India) for tests that meet the specifications outlined in the TPPs. The yearly PAM was estimated for the most likely application of each TPP. In 2020 the PAM for all 4 countries together was estimated to be (1) 12M tests/year with a value of 48M-71M USD for a sputum smear-replacement test; (2) 16M tests/year with a value of 65M-97M USD for a biomarker test; (3) 18M tests/year with a value of 18M-35M USD for a triage test; (4) 12M tests/year with a value of 59M-2238M USD for a tuberculosis detection plus drug susceptibility test (DST) all-in-one or 1.5M tests/year for a DST that follows a positive tuberculosis detection test with a corresponding value of 75M-121M for both tuberculosis detection and DST. Although there is a considerable potential market for novel tuberculosis diagnostics that fit the specification of the TPPs in the 4 high-burden countries, the actual market for an individual product remains uncertain.
Sigma-Aldrich
meso-Tetraphenylporphyrin, BioReagent, suitable for fluorescence, ≥99.0% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Rifampicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC), powder or crystals
Pricing and availability is not currently available.
Sigma-Aldrich
Rifampicin, ≥95% (HPLC), powder or crystals
Pricing and availability is not currently available.