Merck
  • Absorbance detector for capillary electrophoresis based on light-emitting diodes and photodiodes for the deep-ultraviolet range.
Absorbance detector for capillary electrophoresis based on light-emitting diodes and photodiodes for the deep-ultraviolet range.
Journal of chromatography. A (2015-06-21)
Duy Anh Bui, Peter C Hauser
ABSTRACT
A new absorbance detector for capillary electrophoresis featuring relatively high intensity light-emitting diodes as radiation sources and photodiodes for the deep-UV range was developed. The direct relationship of absorbance values and concentrations was obtained by emulating Lambert-Beer's law with the application of a beam splitter to obtain a reference signal and a log-ratio amplifier circuitry. The performance of the cell was investigated at 255 nm with the detection of sulfanilic, 4-nitrobenzoic, 4-hydroxybenzoic and 4-aminobenzoic acid and the indirect detection of acetate, propionate, butyrate and caproate using benzoate as the displacement dye molecule. Vanillic acid, L-tyrosine and DL-tryptophan as well as the sulfonamides sulfamerazine, sulfathiazole and sulfamethazine were determined at 280 nm. Good linearities over 3 orders of magnitude were obtained. The noise level recorded was as low as 50 μAU and the drift typically <200 μAU/5 min.
Sigma-Aldrich
Sodium propionate, ≥99.0%
Sigma-Aldrich
Sodium propionate, ≥99.0%, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Sodium propionate, meets analytical specification of NF, 99-100.5% (ex dried subst.)
Sigma-Aldrich
Sulfanilic acid, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium propionate, FCC
Sigma-Aldrich
Sulfanilic acid, JIS special grade, 99.0-100.5%
Sigma-Aldrich
Sulfanilic acid, ACS reagent, 99%
Sigma-Aldrich
Sodium butyrate, 98%
Sigma-Aldrich
Sodium butyrate, ≥98.5% (GC)
Sigma-Aldrich
Sodium butyrate, Vetec, reagent grade, 99%