Merck
  • Direct identification of on-bead peptides using surface-enhanced Raman spectroscopic barcoding system for high-throughput bioanalysis.
Direct identification of on-bead peptides using surface-enhanced Raman spectroscopic barcoding system for high-throughput bioanalysis.
Scientific reports (2015-05-29)
Homan Kang, Sinyoung Jeong, Yul Koh, Myeong Geun Cha, Jin-Kyoung Yang, San Kyeong, Jaehi Kim, Seon-Yeong Kwak, Hye-Jin Chang, Hyunmi Lee, Cheolhwan Jeong, Jong-Ho Kim, Bong-Hyun Jun, Yong-Kweon Kim, Dae Hong Jeong, Yoon-Sik Lee
ABSTRACT
Recently, preparation and screening of compound libraries remain one of the most challenging tasks in drug discovery, biomarker detection, and biomolecular profiling processes. So far, several distinct encoding/decoding methods such as chemical encoding, graphical encoding, and optical encoding have been reported to identify those libraries. In this paper, a simple and efficient surface-enhanced Raman spectroscopic (SERS) barcoding method using highly sensitive SERS nanoparticles (SERS ID) is presented. The 44 kinds of SERS IDs were able to generate simple codes and could possibly generate more than one million kinds of codes by incorporating combinations of different SERS IDs. The barcoding method exhibited high stability and reliability under bioassay conditions. The SERS ID encoding based screening platform can identify the peptide ligand on the bead and also quantify its binding affinity for specific protein. We believe that our SERS barcoding technology is a promising method in the screening of one-bead-one-compound (OBOC) libraries for drug discovery.
Sigma-Aldrich
Phenyl isothiocyanate, for HPLC derivatization, the detection of alcohols and amines, ≥99.0%
Sigma-Aldrich
Ethylene glycol, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Thiophenol, ≥99%
Sigma-Aldrich
2-Mercaptopyrimidine, 98%
Sigma-Aldrich
Dibenzyl disulfide, 98%
Sigma-Aldrich
3-Cyanobenzoic acid, 98%
Sigma-Aldrich
4-Chlorothiophenol, 97%
Sigma-Aldrich
4-Aminothiophenol, ≥97.0%
Sigma-Aldrich
Phenyl isothiocyanate, 99%, for protein sequencing
Sigma-Aldrich
2-Mercapto-1-methylimidazole, ≥99%
Sigma-Aldrich
2-Thiazoline-2-thiol, 98%
Sigma-Aldrich
3,5-Dimethylbenzenethiol, 90%
Sigma-Aldrich
4,4′−Dipyridyl, 98%
Sigma-Aldrich
Phenyl isothiocyanate, reagent grade, 98%
Sigma-Aldrich
3,4-Dimethylbenzenethiol, 98%
Sigma-Aldrich
Thiophenol, 97%
Sigma-Aldrich
2-Thiouracil, ≥99%
Sigma-Aldrich
Phenyl isothiocyanate, Sigma Grade, 8.36 M, suitable for solid phase protein sequencing analysis, ≥99% (GC), liquid
Supelco
Ethylene glycol solution, NMR reference standard, 80% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
4-Mercaptobenzoic acid, 99%
Sigma-Aldrich
4-Mercaptobenzoic acid, technical grade, 90%
Sigma-Aldrich
Ethylene glycol, SAJ first grade, ≥99.0%
Sigma-Aldrich
Ethylene glycol, JIS special grade, ≥99.5%
Sigma-Aldrich
Ethylene glycol, anhydrous, 99.8%
Sigma-Aldrich
Ethylene glycol 5 M solution
Sigma-Aldrich
4-Bromothiophenol, 95%
Sigma-Aldrich
2-Mercaptobenzimidazole, 98%
Sigma-Aldrich
N,N-Diisopropylethylamine, purified by redistillation, 99.5%
Sigma-Aldrich
2-Bromothiophenol, 97%
Sigma-Aldrich
4-Fluorothiophenol, 98%