Merck
  • HPLC-UV method validation for the identification and quantification of bioactive amines in commercial eggs.
HPLC-UV method validation for the identification and quantification of bioactive amines in commercial eggs.
Talanta (2015-05-25)
Tadeu Chaves de Figueiredo, Débora Cristina Sampaio de Assis, Liliane Denize Miranda Menezes, Guilherme Resende da Silva, Isabela Pereira Lanza, Luiz Guilherme Dias Heneine, Silvana de Vasconcelos Cançado
ABSTRACT
A quantitative and confirmatory high-performance liquid chromatography with ultraviolet detection (HPLC-UV) method for the determination of bioactive amines in the albumen and yolk of commercial eggs was developed, optimized and validated by analyte extraction with trichloroacetic acid and pre-column derivatization with dansyl chloride. Phenylethylamine, putrescine, cadaverine, histamine, tyramine, spermidine and spermine standards were used to evaluate the following performance parameters: limit of detection (LoD), limit of quantification (LoQ), selectivity, linearity, precision, recovery and ruggedness. The LoD of the method was defined from 0.2 to 0.3 mg kg(-1) for the yolk matrix and from 0.2 to 0.4 mg kg(-1) for the albumen matrix; the LoQ was from 0.7 to 1.0 mg kg(-1) for the yolk matrix and from 0.7 to 1.1 mg kg(-1) for the albumen matrix. The validated method exhibited excellent selectivity and separation of all amines with coefficients of determination higher than 0.99. The obtained recovery values were from 90.5% to 108.3%, and the relative standard deviation (RSD) was lower than 10% under repeatability conditions for the studied analytes. The performance parameters show the validated method to be adequate for the determination of bioactive amines in egg albumen and yolk.
Sigma-Aldrich
Acetonitrile, for residue analysis, JIS 5000
Sigma-Aldrich
Acetonitrile, for chromatography
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 1000
Sigma-Aldrich
Toluene, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 300
Sigma-Aldrich
Toluene, JIS 300, for residue analysis, ≥99.8%
Sigma-Aldrich
Toluene, JIS special grade, ≥99.5%
Sigma-Aldrich
Acetonitrile, JIS special grade, ≥99.5%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, 200 μm particle size
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N
Sigma-Aldrich
Poly(tetrafluoroethylene), beads
Sigma-Aldrich
Spermidine trihydrochloride, ≥98% (TLC)
Sigma-Aldrich
Spermidine trihydrochloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Nitrogen, ≥99.998%
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, >40 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder (free-flowing), 1 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, 35 μm particle size
SAFC
Spermidine trihydrochloride, ≥99.0% (AT)
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Supelco
Toluene, HPLC grade, 99.8%
Sigma-Aldrich
Poly(tetrafluoroethylene), powder (free-flowing), ≤12 μm particle size
Sigma-Aldrich
Toluene, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, ≥350 μm particle size
Sigma-Aldrich
Toluene, JIS 1000, for residue analysis, ≥99.8%