Merck
  • Deletion of Stat3 in hematopoietic cells enhances thrombocytosis and shortens survival in a JAK2-V617F mouse model of MPN.
Deletion of Stat3 in hematopoietic cells enhances thrombocytosis and shortens survival in a JAK2-V617F mouse model of MPN.
Blood (2015-01-18)
Jean Grisouard, Takafumi Shimizu, Adrian Duek, Lucia Kubovcakova, Hui Hao-Shen, Stephan Dirnhofer, Radek C Skoda
ABSTRACT
The acquired somatic JAK2-V617F mutation is present in >80% of patients with myeloproliferative neoplasms (MPNs). Stat3 plays a role in hematopoietic homeostasis and might influence the JAK2-V617F-driven MPN phenotype. We crossed our transgenic SclCre;V617F mice with a conditional Stat3 knockout strain and performed bone marrow transplantations into lethally irradiated recipient mice. The deletion of Stat3 increased the platelet numbers in SclCre;V617F;Stat3(fl/fl) mice compared with SclCre;V617F;Stat3(fl/+) or SclCre;V617F;Stat3(+/+) mice. Stat3 deletion also normalized JAK2-V617F-induced neutrophilia. Megakaryocyte progenitors were elevated, especially in the spleen, and a slight increase in myelofibrosis was noted. We observed increased mRNA expression levels of Stat1 and Stat1 target genes and augmented phosphorylation of Stat1 protein in bone marrow and spleen of JAK2-V617F mice after Stat3 deletion. The survival of Stat3-deficient mice expressing JAK2-V617F was reduced. Inflammatory bowel disease, previously associated with shortened survival of Stat3-deficient mice, was less prominent in the bone marrow transplantation setting, possibly by limiting deletion of Stat3 to hematopoietic tissues only. In conclusion, deletion of Stat3 in hematopoietic cells from JAK2-V617F mice did not ameliorate the course of MPN, but rather enhanced thrombocytosis and shortened the overall survival.
Sigma-Aldrich
Sucrose, SAJ first grade
Sigma-Aldrich
Sucrose, JIS special grade
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, Grade I, ≥99% (GC), suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
S3I-201, ≥97% (HPLC)
Sigma-Aldrich
Tamoxifen, ≥99%