Merck
  • CXCL5/CXCR2 axis promotes bladder cancer cell migration and invasion by activating PI3K/AKT-induced upregulation of MMP2/MMP9.
CXCL5/CXCR2 axis promotes bladder cancer cell migration and invasion by activating PI3K/AKT-induced upregulation of MMP2/MMP9.
International journal of oncology (2015-06-11)
Ye Gao, Zhenfeng Guan, Jiaqi Chen, Hongjun Xie, Zhao Yang, Jinhai Fan, Xinyang Wang, Lei Li
ABSTRACT
Bladder cancer (BCa) is the most common malignant disease of the urinary tract system, yet the etiology is still poorly understood. Clinically, the majority of BCa patients progress to invasive disease at the final stage, leading to death. Previous investigations have demonstrated that matrix metal-loproteinases (MMPs) play irreplaceable roles in tumor cell extravasation and implantation. In addition, increasing numbers of reports provide evidence that MMPs, especially MMP2 and MMP9 are monitored by various signal transduction pathways targeting tumor metastasis. Seed-and-soil theory has called to attention the importance of the tumor microenvironment in disease progression. To that end, we previously reported the key role of hypoxia in BCa progression. Herein, we report the role of chemokines, specifically CXCL5, is involved in BCa development. Though it has been reported that CXCL5 promotes BCa metastasis and progression, the exact mechanisms are still unknown, necessitating the need for further investigation into the role of CXCL5 in BCa. In this study, IHC staining of BCa tumor sections showed elevated expression of CXCL5 in BCa, which correlated with disease stage. Our mechanistic studies show that CXCL5 contributes to BCa migration and invasion by binding to its receptor, CXCR2, leading to the upregulation of MMP2/MMP9 by activating PI3K/AKT signaling. This study offers vital evidence of how CXCL5 promotes BCa metastasis, and thus may potentially be used as a therapeutic target against BCa.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, SAJ special grade, ≥97.0%
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Mechlorethamine hydrochloride, 98%
Sigma-Aldrich
Human ENA78 / CXCL5 ELISA Kit, for serum, plasma, cell culture supernatants and urine
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Akt1
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
Hematoxylin, certified by the Biological Stain Commission
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
MISSION® esiRNA, targeting human AKT1, RP11-982M15.2
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)
SAFC
Sodium deoxycholate