Merck
  • Effects of normal and high circulating concentrations of activin A on vascular endothelial cell functions and vasoactive factor production.
Effects of normal and high circulating concentrations of activin A on vascular endothelial cell functions and vasoactive factor production.
Pregnancy hypertension (2015-11-26)
Hannah E J Yong, Padma Murthi, May H Wong, Bill Kalionis, Judith E Cartwright, Shaun P Brennecke, Rosemary J Keogh
ABSTRACT
Activin A, a TGFβ family member, circulates in the maternal blood at increasing concentrations throughout gestation during a healthy pregnancy. The circulating concentration of activin A is further increased in pre-eclampsia (PE), a hypertensive disorder of pregnancy that is marked by systemic maternal vascular endothelial cell dysfunction. The effect of increasing activin A concentrations on the maternal vascular endothelium is unknown. The study aim was to investigate the effect of physiological and pathological activin A concentrations observed in normotensive and PE pregnancies respectively, on vascular endothelial cell function. Immunostaining demonstrated the presence of the activin A receptor, ACVR2A, in SGHEC-7 cells used to model the vascular endothelium. SGHEC-7 cells were treated with activin A concentrations representative of concentrations throughout gestation in normotensive (0-10ng/ml) and PE (50ng/ml) pregnancies. xCELLigence functional assays revealed that normotensive activin A concentrations increased SGHEC-7 proliferation and migration, which was inhibited by PE concentrations. Additionally, fluorescence based assays showed that PE concentrations increased endothelial permeability. None of the tested activin A concentrations affected cell apoptosis. PE concentrations also resulted in an imbalance of the vasoactive factors eNOS, PTGIS and EDN1, as determined by quantitative real-time polymerase chain reaction and enzyme-linked immunosorbent assays. Compared with normotensive activin A concentrations, the higher PE activin A concentrations resulted in abnormal endothelial functions, which may contribute to the systemic maternal vascular endothelial cell dysfunction observed in the disorder.
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., suitable for cell culture, ≥95%
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ≥95% (HPLC)
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ~98% (HPLC)
Sigma-Aldrich
Activin A human, recombinant, expressed in HEK 293 cells, HumanKine®, suitable for cell culture
Sigma-Aldrich
Activin A human, recombinant, expressed in E. coli, ≥97% (SDS-PAGE), ≥97% (HPLC), suitable for cell culture
Sigma-Aldrich
Activin A human, ≥95% (SDS-PAGE), recombinant, expressed in baculovirus infected insect cells, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Activin A active human, Animal-component free, recombinant, expressed in Nicotiana, >97% (SDS-PAGE)