Merck
  • GABA from reactive astrocytes impairs memory in mouse models of Alzheimer's disease.
GABA from reactive astrocytes impairs memory in mouse models of Alzheimer's disease.
Nature medicine (2014-06-30)
Seonmi Jo, Oleg Yarishkin, Yu Jin Hwang, Ye Eun Chun, Mijeong Park, Dong Ho Woo, Jin Young Bae, Taekeun Kim, Jaekwang Lee, Heejung Chun, Hyun Jung Park, Da Yong Lee, Jinpyo Hong, Hye Yun Kim, Soo-Jin Oh, Seung Ju Park, Hyo Lee, Bo-Eun Yoon, YoungSoo Kim, Yong Jeong, Insop Shim, Yong Chul Bae, Jeiwon Cho, Neil W Kowall, Hoon Ryu, Eunmi Hwang, Daesoo Kim, C Justin Lee
ABSTRACT
In Alzheimer's disease (AD), memory impairment is the most prominent feature that afflicts patients and their families. Although reactive astrocytes have been observed around amyloid plaques since the disease was first described, their role in memory impairment has been poorly understood. Here, we show that reactive astrocytes aberrantly and abundantly produce the inhibitory gliotransmitter GABA by monoamine oxidase-B (Maob) and abnormally release GABA through the bestrophin 1 channel. In the dentate gyrus of mouse models of AD, the released GABA reduces spike probability of granule cells by acting on presynaptic GABA receptors. Suppressing GABA production or release from reactive astrocytes fully restores the impaired spike probability, synaptic plasticity, and learning and memory in the mice. In the postmortem brain of individuals with AD, astrocytic GABA and MAOB are significantly upregulated. We propose that selective inhibition of astrocytic GABA synthesis or release may serve as an effective therapeutic strategy for treating memory impairment in AD.
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein Antibody, Chemicon®, from chicken
Sigma-Aldrich
Anti-Actin Antibody, clone C4, clone C4, Chemicon®, from mouse
Sigma-Aldrich
Anti-GABA Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-GAD67 Antibody, clone 1G10.2, clone 1G10.2, Chemicon®, from mouse
Sigma-Aldrich
Anti-GABA Transporter-1 Antibody, CT (a.a. 588-599), Chemicon®, from rabbit
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse