Merck
  • Nabothian cyst a predominant cause of false-positive iodine uptake in uterus: comparison of SPECT/CT and pelvic MRI.
Nabothian cyst a predominant cause of false-positive iodine uptake in uterus: comparison of SPECT/CT and pelvic MRI.
Clinical nuclear medicine (2014-07-01)
Takuro Isoda, Shingo BaBa, Yasuhiro Maruoka, Yoshiyuki Kitamura, Akihiro Nishie, Masayuki Sasaki, Hiroshi Honda
ABSTRACT
Many cases of unexpected radioiodine uptake have been reported, including physiological uptake in healthy tissue and in both benign and malignant nonthyroidal lesions. However, iodine uptake in the uterus has not been well assessed. In this article, we systemically analyzed iodine uptake in the uterus and identified the site in which uptake occurred. Seventy-six female patients who underwent SPECT/CT from neck to pelvis were included in this study; 5 patients with uterine uptake received pelvic MRI. Iodine uptake in the uterus was diagnosed with SPECT/CT and compared with the findings of the MRI. Serum TSH level, administered dose of iodine-131 and number of radioiodine therapies were compared between the 2 groups with and without iodine uptake in the uterus. Twenty (26.3%) of 76 patients showed iodine uptake in the uterine cervix on SPECT/CT. It was difficult to distinguish the uptake in the uterus from that in the urinary bladder or rectum using only planar images. The patients with the uterine uptake were younger than those without (median age, 46.5 vs. 62, P = 0.011). There were no significant differences in serum TSH level, administered dose of iodine-131, or number of radioiodine therapies between the 2 groups. In all instances of the 5 patients who underwent pelvic MRI, iodine uptake in the uteri was located in nabothian cysts. Iodine uptake in the uterus is frequently found if pelvic SPECT/CT is performed. Nabothian cysts are a predominant cause of iodine uptake in the uterine cervix.
Sigma-Aldrich
Sodium iodide, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium iodide, 99.999% trace metals basis
Sigma-Aldrich
Sodium iodide, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium iodide, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium iodide, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium iodide, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium iodide, puriss., meets analytical specification of Ph. Eur., BP, USP, 99-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium iodide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%