Merck
  • A facile strategy to functionalize gold nanorods with polycation brushes for biomedical applications.
A facile strategy to functionalize gold nanorods with polycation brushes for biomedical applications.
Acta biomaterialia (2014-05-13)
Peng Yan, Nana Zhao, Hao Hu, Xinyi Lin, Fusheng Liu, Fu-Jian Xu
ABSTRACT
The fabrication of highly efficient nonviral gene carriers with low cytotoxicity remains a challenge in gene therapy. This paper reports a facile strategy to combine the advantages of gold nanorods (Au NRs) and polycations through surface functionalization. Different Au NR carriers with a controlled amount of poly(2-(N,N-dimethyl amino)ethyl methacrylate) (PDAEMA) brushes could be readily synthesized via surface-initiated atom transfer radical polymerization to achieve optimized nanohybrids for gene transfection. The obtained gene carriers demonstrate much higher gene transfection efficiency and lower cytotoxicity compared with polyethylenimine (∼25kDa, gold standard of nonviral gene vector) in both COS7 and HepG2 cell lines. In addition, the potential of the PDMAEMA-grafted Au NR carriers to be utilized as a computed tomography contrast agent for the imaging of cancer cells has also been investigated. This strategy may realize the gene therapy and real-time imaging within one nanostructure and facilitate biomedical applications.
Sigma-Aldrich
Diethylenetriamine, SAJ first grade, ≥98.0%
Sigma-Aldrich
Triethylamine, SAJ first grade, ≥98.0%
Sigma-Aldrich
N,N-Dimethylformamide, SAJ first grade, ≥99.0%
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
N,N-Dimethylformamide, suitable for HPLC, ≥99.9%
Sigma-Aldrich
VenPure® SF, powder
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
Copper(I) bromide, AnhydroBeads, -10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Copper(I) bromide, 99.999% trace metals basis
Sigma-Aldrich
N,N-Dimethylformamide, JIS special grade, ≥99.5%
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
N,N-Dimethylformamide, for molecular biology, ≥99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Supelco
N,N-Dimethylformamide, analytical standard
Sigma-Aldrich
2-Bromo-2-methylpropionic acid, 98%
Sigma-Aldrich
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%