Merck
  • Fluvastatin attenuates hepatic steatosis-induced fibrogenesis in rats through inhibiting paracrine effect of hepatocyte on hepatic stellate cells.
Fluvastatin attenuates hepatic steatosis-induced fibrogenesis in rats through inhibiting paracrine effect of hepatocyte on hepatic stellate cells.
BMC gastroenterology (2015-04-19)
Lee-Won Chong, Yi-Chao Hsu, Ting-Fang Lee, Yun Lin, Yung-Tsung Chiu, Kuo-Ching Yang, Jaw-Ching Wu, Yi-Tsau Huang
ABSTRACT
Non-alcoholic steatohepatitis (NASH) is associated with hepatic fibrogenesis. Despite well-known cholesterol-lowering action of statins, their mechanisms against NASH-mediated fibrogenesis remain unclear. This study aimed at investigating the in vitro and in vivo anti-fibrotic properties of fluvastatin (Flu). Palmitate (PA)-induced changes in intracellular hydrogen peroxide levels in primary rat hepatocytes (PRHs) and human hepatoma cell line (HepG2) were quantified by dichlorofluorescein diacetate (DCF-DA) dye assay, whereas changes in expressions of NADPH oxidase gp91 (phox) subunit, α-smooth muscle actin (α-SMA), and NFκB p65 nuclear translocation were quantified with Western blotting. Quantitative real-time polymerase chain reaction (q-PCR) was used to investigate mRNA expressions of pro-inflammatory genes (ICAM-1, IL-6, TNF-α). Conditioned medium (CM) from PA-treated PRHs was applied to cultured rat hepatic stellate cell line, HSC-T6, with or without Flu-pretreatment for 2 h. Pro-fibrogenic gene expressions (COL1, TIMP-1, TGF-β1, α-SMA) and protein expression of α-SMA were analyzed. In vivo study using choline-deficient L-amino acid defined (CDAA) diet-induced rat NASH model was performed by randomly assigning Wistar rats (n = 28) to normal controls (n = 4), CDAA diet with vehicles, and CDAA diet with Flu (5 mg/kg or 10 mg/kg) (n = 8 each) through gavage for 4 or 8 weeks. Livers were harvested for histological, Western blot (α-SMA), and q-PCR analyses for expressions of pro-inflammatory (IL-6, iNOS, ICAM-1) and pro-fibrogenic (Col1, α-SMA, TIMP-1) genes. In vitro, Flu (1-20 μM) inhibited PA-induced free-radical production, gp91 (phox) expression, and NFκB p65 translocation in HepG2 and PRHs, while CM-induced α-SMA protein expression and pro-fibrogenic gene expressions in HSC-T6 were suppressed in Flu-pretreated cells compared to those without pretreatment. Moreover, α-SMA protein expression was significantly decreased in HSC-T6 cultured with CM from PA-Flu-treated PRHs compared to those cultured with CM from PA-treated PRHs. Flu also reduced steatosis and fibrosis scores, α-SMA protein expression, mRNA expression of pro-inflammatory and pro-fibrogenic genes in livers of CDAA rats. We demonstrated PA-induced HSC activation through paracrine effect of hepatocyte in vitro that was significantly suppressed by pre-treating HSC with Flu. In vivo, Flu alleviated steatosis-induced HSC activation and hepatic fibrogenesis through mitigating inflammation and oxidative stress, suggesting possible therapeutic role of Flu against NASH.
Product Description
Fluvastatin sodium, European Pharmacopoeia (EP) Reference Standard
USP
Fluvastatin sodium, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Anti-β-Catenin Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-gp91-phox Antibody, Upstate®, from rabbit
Sigma-Aldrich
Sodium palmitate, ≥98.5%
Fluvastatin for system suitability, European Pharmacopoeia (EP) Reference Standard