Merck
  • Paper spray and extraction spray mass spectrometry for the direct and simultaneous quantification of eight drugs of abuse in whole blood.
Paper spray and extraction spray mass spectrometry for the direct and simultaneous quantification of eight drugs of abuse in whole blood.
Analytical chemistry (2014-06-28)
Ryan D Espy, Sebastiaan Frans Teunissen, Nicholas E Manicke, Yue Ren, Zheng Ouyang, Arian van Asten, R Graham Cooks
ABSTRACT
Determination of eight drugs of abuse in blood has been performed using paper spray or extraction spray mass spectrometry in under 2 min with minimal sample preparation. A method has been optimized for quantification of amphetamine, methamphetamine, 3,4-methylenedioxyamphetamine (MDA), 3,4-methylenedioxy-N-methylamphetamine (MDMA), 3,4-methylenedioxy-N-ethylamphetamine (MDEA), morphine, cocaine, and Δ9-tetrahydrocannabinol (THC) from a single blood spot. Sample to sample variations of 1-5% relative standard deviation were achieved using stable isotope-labeled internal standards and tandem mass spectrometry. Limits of detection for all drugs were below typical physiological and toxicological levels. Paper spray and extraction spray each used less than 10 μL of whole blood. These methods exhibit the potential for performing rapid and high-throughput assays for selective on-site multicompound quantitative screening of illicit drugs.
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Sodium fluoride 0.5 M solution
Sigma-Aldrich
Sodium fluoride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Supelco
Fluoride ion solution for ISE, 0.1 M F-, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Sodium fluoride, JIS special grade, ≥99.0%
Supelco
Sodium Fluoride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium fluoride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium fluoride, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%