Merck
  • Stimulation of bone growth following zinc incorporation into biomaterials.
Stimulation of bone growth following zinc incorporation into biomaterials.
Biomaterials (2014-05-28)
Yuqin Qiao, Wenjie Zhang, Peng Tian, Fanhao Meng, Hongqin Zhu, Xinquan Jiang, Xuanyong Liu, Paul K Chu
ABSTRACT
Rapid development of zinc biology has broadened the applications of Zn-incorporated biomaterials to tissue engineering but also raised concerns about the long-term safety of released Zn(2+) ions. Clinical success hinges on the amount of incorporated zinc and subsequent optimized release sufficient to stimulate osseointegration. In this study, zinc is incorporated into the sub-surface of TiO2 coatings by plasma immersion ion implantation and deposition (PIII&D). The Zn-implanted coatings show significant improvement compared to the "bulk-doped" coatings prepared by plasma electrolyte oxidation in terms of osteogenesis in vitro and in vivo. Molecular and cellular osteogenic activities demonstrate that rBMSCs cultured on the Zn-implanted coatings have higher ALP activity and up-regulated osteogenic-related genes (OCN, Col-I, ALP, Runx2) compared to the bulk-doped Zn coatings and controls. In vivo osseointegration studies conducted for 12 weeks on the rat model show early-stage new bone formation and the bone contact ratio (12 week) on the Zn-implanted coating is larger. The ZnT1 and ZIP1 gene expression studies demonstrate that the Zn-implanted coatings can better stimulate bone growth with reduced Zn release than those doped with zinc throughout the coatings.
Product Description
Supelco
Tetracycline hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Tetracycline hydrochloride, VETRANAL®, analytical standard
Sigma-Aldrich
Tetracycline hydrochloride, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Tetracycline hydrochloride, meets USP testing specifications
Sigma-Aldrich
Tetracycline hydrochloride, ≥95% (European Pharmacopoeia HPLC assay)
Sigma-Aldrich
Tetracycline hydrochloride, powder
Sigma-Aldrich
Calcein, Used for the fluorometric determination of calcium and EDTA titration of calcium in the presence of magnesium.
Tetracycline hydrochloride, European Pharmacopoeia (EP) Reference Standard
USP
Tetracycline hydrochloride, United States Pharmacopeia (USP) Reference Standard