Merck
  • KCa3.1 modulates neuroblast migration along the rostral migratory stream (RMS) in vivo.
KCa3.1 modulates neuroblast migration along the rostral migratory stream (RMS) in vivo.
Cerebral cortex (New York, N.Y. : 1991) (2013-04-16)
Kathryn L Turner, Harald Sontheimer
ABSTRACT
From the subventricular zone (SVZ), neuronal precursor cells (NPCs), called neuroblasts, migrate through the rostral migratory stream (RMS) to become interneurons in the olfactory bulb (OB). Ion channels regulate neuronal migration during development, yet their role in migration through the adult RMS is unknown. To address this question, we utilized Nestin-CreER(T2)/R26R-YFP mice to fluorescently label neuroblasts in the adult. Patch-clamp recordings from neuroblasts reveal K(+) currents that are sensitive to intracellular Ca(2+) levels and blocked by clotrimazole and TRAM-34, inhibitors of intermediate conductance Ca(2+)-activated K(+) (KCa3.1) channels. Immunolabeling and electrophysiology show KCa3.1 expression restricted to neuroblasts in the SVZ and RMS, but absent in OB neurons. Time-lapse confocal microscopy in situ showed inhibiting KCa3.1 prolonged the stationary phase of neuroblasts' saltatory migration, reducing migration speed by over 50%. Both migration and KCa3.1 currents could also be inhibited by blocking Ca(2+) influx via transient receptor potential (TRP) channels, which, together with positive immunostaining for transient receptor potential canonical 1 (TRPC1), suggest that TRP channels are an important Ca(2+) source modulating KCa3.1 activity. Finally, injecting TRAM-34 into Nestin-CreER(T2)/R26R-YFP mice significantly reduced the number of neuroblasts that reached the OB, suggesting an important role for KCa3.1 in vivo. These studies describe a previously unrecognized protein in migration of adult NPCs.
Product Description
Supelco
Clotrimazole, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Apamin, synthetic, ≥97% (HPLC)
Sigma-Aldrich
Apamin, from bee venom, ≥95% (HPLC)
Sigma-Aldrich
Clotrimazole
Sigma-Aldrich
2-Aminoethyl diphenylborinate, 97%
Sigma-Aldrich
Tamoxifen, ≥99%
USP
Clotrimazole, United States Pharmacopeia (USP) Reference Standard
Clotrimazole, European Pharmacopoeia (EP) Reference Standard
Supelco
Tamoxifen, analytical standard
Clotrimazole for peak identification, European Pharmacopoeia (EP) Reference Standard